Skip to Content
Merck

SML3619

Linagliptin

≥98% (HPLC), dipeptidyl peptidase IV inhibitor, powder

Synonym(s):

(R)-8-(3-Amino-piperidin-1-yl)-7-but-2-ynyl-3-methyl-1-(4-methyl-quinazolin-2-ylmethyl)-3,7-dihydro-purine-2,6-dione, 8-(3-(R)-Aminopiperidin-1-yl)-7-but-2-ynyl-3-methyl-1-(4-methyl-quinazolin-2-ylmethyl)-3,7-dihydropurine-2,6-dione, BI 1356, BI-1356, BI1356, Ondero

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H28N8O2
CAS Number:
Molecular Weight:
472.54
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Linagliptin, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N5(C[C@@H](CCC5)N)c1[n](c2c([n]([c]([n]([c]2=O)Cc3nc4c(c(n3)C)cccc4)=O)C)n1)CC#CC

InChI

1S/C25H28N8O2/c1-4-5-13-32-21-22(29-24(32)31-12-8-9-17(26)14-31)30(3)25(35)33(23(21)34)15-20-27-16(2)18-10-6-7-11-19(18)28-20/h6-7,10-11,17H,8-9,12-15,26H2,1-3H3/t17-/m1/s1

InChI key

LTXREWYXXSTFRX-QGZVFWFLSA-N

Biochem/physiol Actions

Linagliptin is an orally active, competitive, potent and selective dipeptidyl peptidase IV (DPP4; DPP-IV) inhibitor (IC50 = 1 nM vs. 19 nM/sitagliptin, 24 nM/alogliptin, 50 nM/saxagliptin, 62 nM/vildagliptin) with no inhibitory potency against DPP-2/8/9 (IC50 >10 µM). Sitagliptin improves glucose tolerance and exhibits efficacy in diabetic rodent models in vivo (1-10 mg/kg via daily p.o. in mice and rats).
Orally active, competitive, potent and selective dipeptidyl peptidase IV (DPP-IV; DPP4) inhibitor that improves glucose tolerance in vivo.


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library