Skip to Content
Merck
All Photos(1)

Key Documents

SML3600

Sigma-Aldrich

AZ10606120 dihydrochloride

≥98% (HPLC)

Synonym(s):

2-(1-Adamantyl)-N-[2-({2-[(2-hydroxyethyl)amino]ethyl}amino)quinolin-5-yl]acetamide dihydrochloride, AZ 10606120 dihydrochloride, AZ-10606120 dihydrochloride, Compound-17 dihydrochloride, N-[2-({2-[(2-Hydroxyethyl)amino]ethyl}amino)-5-quinolinyl]-2-tricyclo[3.3.1.13,7]dec-1-ylacetamidee dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34N4O2·2HCl
CAS Number:
Molecular Weight:
495.48
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.04

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated
protect from light
under inert gas

color

white to beige

solubility

H2O: 2 mg/mL, clear (Warmed)

storage temp.

room temp

SMILES string

O=C(NC1=CC=CC2=C1C=CC(NCCNCCO)=N2)CC3(C[C@H]4C5)C[C@@H](C4)C[C@@H]5C3.[2HCl]

Biochem/physiol Actions

AZ-10606120 is a potent purinoceptor P2X7 negative allosteric modulator that antagonizes ATP-dependent P2X7 activation (IC50 against ATP-induced ethidium bromide uptake = 2.3 nM and 24 nM, respectively, using human or rat P2X7 HEK293 transfectants; [ATP] = EC50) in a non-competitive and slowly reversible manner via affinity interaction at an allosteric pocket (human/rat P2X7 KD = 1.4 nM/19 nM) distinct from the KN-62- or ATP-binding site. A useful tool for probing P2X7-mediated physiological and pathological processes both in cultures (0.1-10 µM) and in vivo (e.g. 5 mg/kg i.p., 300 nM i.m., or 3 µg i.c.v.; in mice).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service