Skip to Content
Merck

SML3365

MZ1

≥98% (HPLC)

Synonym(s):

(2S,4R)-1-((2S)-2-tert-Butyl-17-((6S)-4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-4,16-dioxo-6,9,12-trioxa-3,15-diazaheptadecane)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, (2S,4R)-1-((S)-1-((S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-17,17-dimethyl-2,14-dioxo-6,9,12-trioxa-3,15-diazaoctadecanecarbonyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide, (4R)-N-[14-[(6S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl]-1,13-dioxo-3,6,9-trioxa-12-azatetradec-1-yl]-3-methyl-L-valyl-4-hydroxy-N-[[4-(4-methyl-5-thiazolyl)phenyl]methyl]-L-prolinamide, MZ 1, MZ 1 (PROTAC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C49H60ClN9O8S2
CAS Number:
Molecular Weight:
1002.64
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C([C@H]1N(C[C@@H](C1)O)C([C@H](C(C)(C)C)NC(COCCOCCOCCNC(C[C@H]2C3=NN=C(N3C4=C(C(C5=CC=C(C=C5)Cl)=N2)C(C)=C(S4)C)C)=O)=O)=O)NCC6=CC=C(C=C6)C7=C(N=CS7)C

InChI key

PTAMRJLIOCHJMQ-PYNGZGNASA-N

Biochem/physiol Actions

BRD4 degrader
MZ1 is a BRD4 degrader composed of pan-BET selective bromodomain inhibitor and the E3 ubiquitin ligase VHL ligand that induces preferential degradation of BRD4 over BRD2 and BRD3. MZ1exhibits antiproliferative effects in AML colorectal cancer cells lines.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library