Skip to Content
Merck

SML3188

Isavuconazonium sulfate

≥95% (HPLC)

Synonym(s):

(2-{[(1-{1-[(2R,3R)-3-[4-(4-cyanophenyl)-1,3-thiazol-2-yl]-2-(2,5-difluorophenyl)-2-hydroxybutyl]-1H-1,2,4-triazol-4-ium-4-yl}ethoxy)carbonyl](methyl)amino}pyridin-3-yl)methyl N-methylglycinate hydrogen sulfate, BAL 8557-002, BAL8557

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C35H35F2N8O5S · SO4H
CAS Number:
Molecular Weight:
814.84
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(O)([O-])=O.N#CC(C=C1)=CC=C1C2=CSC([C@H](C)[C@](O)(CN3C=[N+](C(C)OC(N(C)C4=NC=CC=C4COC(CNC)=O)=O)C=N3)C5=CC(F)=CC=C5F)=N2

InChI

1S/C35H35F2N8O5S.H2O4S/c1-22(33-42-30(18-51-33)25-9-7-24(15-38)8-10-25)35(48,28-14-27(36)11-12-29(28)37)19-45-21-44(20-41-45)23(2)50-34(47)43(4)32-26(6-5-13-40-32)17-49-31(46)16-39-3;1-5(2,3)4/h5-14,18,20-23,39,48H,16-17,19H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1/t22-,23?,35+;/m0./s1

InChI key

LWXUIUUOMSMZKJ-KLFWAVJMSA-M

Biochem/physiol Actions

Isavuconazonium sulfate (BAL8557) is a prodrug of isavuconazole (BAL 4815), a triazole antifungal drug. Isavuconazole inhibits fungal sterol biosynthesis by inhibition of cytochrome P450 sterol 14-α-demethylase (CYP51), an enzyme in the sterol biosynthesis pathway that leads from lanosterol to ergosterol. Isavuconazonium sulfate has been approved for the treatment of both invasive aspergillosis and invasive mucormycosis.
Triazole antifungal prodrug.


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library