Skip to Content
Merck

SML2930

LH1307

≥97% (HPLC)

Synonym(s):

2,2′-Dimethyl-3,3′-bis[2-methyl-4-(2-acetamidoethylamino)-methyl-5-(3-cyanopyridin-5-yl-methoxy)phenoxymethyl]-1,1′-biphenyl, LH 1307, LH-1307, N,N′-(2,2′-(4,4′-(2,2′-Dimethylbiphenyl-3,3′-diyl)bis(methylene)bis(oxy)bis(2-((5-cyanopyridin-3-yl)methoxy)-5-methyl-4,1-phenylene))bis(methylene)bis(azanediyl)bis(ethane-2,1-diyl))diacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C54H58N8O6
CAS Number:
Molecular Weight:
915.09
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
room temperature
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

storage condition

room temperature

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N#CC1=CC(COC2=C(C=C(C(OCC3=C(C(C4=CC=CC(COC5=CC(OCC6=CN=CC(C#N)=C6)=C(C=C5C)CNCCNC(C)=O)=C4C)=CC=C3)C)=C2)C)CNCCNC(C)=O)=CN=C1

InChI

1S/C54H58N8O6/c1-35-17-47(29-57-13-15-61-39(5)63)53(65-31-43-19-41(23-55)25-59-27-43)21-51(35)67-33-45-9-7-11-49(37(45)3)50-12-8-10-46(38(50)4)34-68-52-22-54(66-32-44-20-42(24-56)26-60-28-44)48(18-36(52)2)30-58-14-16-62-40(6)64/h7-12,17-22,25-28,57-58H,13

InChI key

RNCINFJGGSQEHN-UHFFFAOYSA-N

Biochem/physiol Actions

LH1307 is a potent programmed cell death-1/programmed death-ligand 1 (PD-1 (CD279, PDCD1)/PD-L1 (B7-H1, PDCD1-L1)) protein-protein interaction (PPI) inhibitor (IC50 = 3 nM by HTRF assay) that induces symmetrically arranged PD-L1 homodimer formation by targeting PD-L1 at the PD-1 binding site. LH1307 effectively blocks cell surface PD-1/PD-L1 interaction between PD-1-expressing Jurkat cells and co-cultured PD-L1-expressing U2OS cells (SHP-1 recruitment IC50 = 79 nM) or co-cultured PD-L1-expressing aAPC/CHO-K1 cells (NFAT signaling restoration EC50 = 763 nM).
Potent programmed cell death-1 (PD-1, CD279, PDCD1)/PD-1 ligand (PD-L1, B7-H1, PDCD1-L1) protein-protein interaction (PPI) inhibitor.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library