Skip to Content
Merck

SML2765

Glutor

≥98% (HPLC), glucose uptake inhibitor, powder

Synonym(s):

(S)-6-Methyl-5-(4-morpholinobenzyl)-4-oxo-2-phenyl-N-(pyridin-3-ylmethyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrazine-6-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C31H32N6O3
Molecular Weight:
536.62
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Glutor, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Glutor is a potent inhibitor of glucose uptake that targets glucose transporters GLUT1, -2, and -3. Inhibition of glucose uptake by glutor leads to upregulation of GLUT3 and GLUT1 expression. Glutor potently and selectively suppresses growth of a variety of cancer cell lines. Glutor and glutaminase inhibitor CB-839 synergistically inhibit tumor cell growth.
potent inhibitor of glucose uptake that targets glucose transporters GLUT-1, -2, and -3


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library