Skip to Content
Merck
All Photos(1)

Key Documents

SML2428

Sigma-Aldrich

Pentabromopseudilin

≥97% (HPLC)

Synonym(s):

Pentabromopseudilin, 2,4-dibromo-6-(3,4,5-tribromo-1H-pyrrol-2-yl)phenol, PBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H4Br5NO
CAS Number:
Molecular Weight:
553.66
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

OC1=C(Br)C=C(Br)C=C1C2=C(Br)C(Br)=C(Br)N2

InChI

1S/C10H4Br5NO/c11-3-1-4(9(17)5(12)2-3)8-6(13)7(14)10(15)16-8/h1-2,16-17H

InChI key

LXMNWKJHYOZUQL-UHFFFAOYSA-N

General description

Pentabromopseudilin (PBP) is a potent antibiotic originally isolated from the marine bacterium Pseudomonas bromoutilis. 2 PBP exhibits cytotoxic, antibacterial and phytotoxic activities, including the inhibition of human 12- and 15-lipoxygenases3 and potent inhibition of myosin-dependent processes. 4-5 PBP was identified as a potent inhibitor of the motor activity of vertebrate myosin V with an IC50 value of 1.2μM. 5 PBP inhibits the ATPase activity of myosin by increasing its affinity for ADP, reducing ATP-binding and hydrolysis rates, and through coupling between actin and nucleotide-binding sites in the motor domain. 4 Recently, it was also demonstrated that PBP suppresses TGF-β activity.

Other Notes

Solubility: Chloroform/dichloromethane (at least 10 mg/ml), methanol/ethanol (at least 1 mg/ml), DMSO (10mM)
Working Stock: PBP can be dissolved in DMSO at concentration of 10mM, solution should be stored in the dark at -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of pentabromo- and pentachloropseudilin, and synthetic analogues--allosteric inhibitors of myosin ATPase.
René Martin et al.
Angewandte Chemie (International ed. in English), 48(43), 8042-8046 (2009-09-10)
Roman Fedorov et al.
Nature structural & molecular biology, 16(1), 80-88 (2009-01-06)
We have identified pentabromopseudilin (PBP) as a potent inhibitor of myosin-dependent processes such as isometric tension development and unloaded shortening velocity. PBP-induced reductions in the rate constants for ATP binding, ATP hydrolysis and ADP dissociation extend the time required per
Matthias Preller et al.
Journal of medicinal chemistry, 54(11), 3675-3685 (2011-05-04)
Myosin activity is crucial for many biological functions. Strong links have been established between changes in the activity of specific myosin isoforms and diseases such as cancer, cardiovascular failure, and disorders of sensory organs and the central nervous system. The
Rachana V Ohri et al.
Organic letters, 7(12), 2501-2504 (2005-06-04)
[reaction: see text] A regioselective synthesis of propargylamines by the coupling of propargyl alcohols with tosylamines and carbamates catalyzed by an air- and moisture-tolerant rhenium-oxo complex is described. The ability to couple functionalized components allows for convergent approaches to nitrogen-containing
P R Burkholder et al.
Applied microbiology, 14(4), 649-653 (1966-07-01)
Evidence is presented for the isolation and identification of bacteria able to synthesize an unusual antibiotic containing five bromine atoms per molecule. The identification and taxonomic position of these bacteria was made by use of a computer in conjunction with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service