Skip to Content
Merck
All Photos(1)

Key Documents

SML2368

Sigma-Aldrich

(Z)-Endoxifen

≥98% (HPLC)

Synonym(s):

4-Hydroxy-N-desmethyl-tamoxifen, Endoxifen, Metabolite BX, N-Desmethyl-4-hydroxytamoxifen, Z-Endoxifen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27NO2
CAS Number:
Molecular Weight:
373.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(CCOc1ccc(cc1)\C(=C(\CC)/c3ccccc3)\c2ccc(cc2)O)C

InChI

1S/C25H27NO2/c1-3-24(19-7-5-4-6-8-19)25(20-9-13-22(27)14-10-20)21-11-15-23(16-12-21)28-18-17-26-2/h4-16,26-27H,3,17-18H2,1-2H3/b25-24-

InChI key

MHJBZVSGOZTKRH-IZHYLOQSSA-N

Application

(Z)-Endoxifen has been used to demonstrate the lack of contribution of subventricular zone and leptomeningeal cells in a study determining the generation of neuroblasts from astrocytes.

Biochem/physiol Actions

(Z)-Endoxifen (endoxifen) is an active tamoxifen metabolite generated via actions of cytochrome P450 (CYP) enzymes CYP3A4/5 and CYP2D6. Endoxifen is more potent than tamoxifen as a selective estrogen receptor modulator (SERM) both in vitro and in vivo with good pharmacokinetics and oral availability (∼80% MCF-7 tumor growth inhibition with 4-8 mg/kg/day endoxifen or 20 mg/kg/day tamoxifen in mice via p.o.). Endoxifen also exhibits 4-fold higher PKC inhibitory potency than tamoxifen and can overcome tamoxifen resistance due to cytochrome CYP2D6 polymorphism.
Orally available, active tamoxifen metabolite with higher estrogen receptor modulator (SERM) activity and anticancer efficacy both in vitro and in vivo.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phyllis Elkins et al.
Journal of pharmaceutical and biomedical analysis, 88, 174-179 (2013-09-24)
(Z)-Endoxifen (4-hydroxy-N-desmethyltamoxifen), an active metabolite generated via actions of CYP3A4/5 and CYP2D6, is a more potent selective estrogen receptor modulator (SERM) than tamoxifen. In the MCF-7 human mammary tumor xenograft model with female athymic mice, (Z)-endoxifen, at an oral dose
Shagufta et al.
European journal of medicinal chemistry, 143, 515-531 (2017-12-06)
Tamoxifen (ICI 46 474), trans-1-(4-β-dimethylaminoethoxyphenyl)-1,2-diphenylbut-1-ene, is the most commonly used drug for the treatment of estrogen receptor positive breast cancer and has been saving lives worldwide for the past four decades. Tamoxifen is considered a pioneering drug due to its ubiquitous
Ateeq Ahmad et al.
Breast cancer research and treatment, 122(2), 579-584 (2010-01-07)
Endoxifen is the key active metabolite of tamoxifen, a widely used breast cancer drug. Orally administered tamoxifen, is extensively metabolized by cytochrome P450 (CYP) enzymes, namely CYP3A4 and CYP2D6, into active metabolites, especially endoxifen. Due to genetic polymorphism of CYP2D6
Todd C Skaar et al.
Clinical pharmacology and therapeutics, 103(5), 755-757 (2018-02-24)
This issue of Clinical Pharmacology & Therapeutics (CPT) includes the Clinical Pharmacogenetics Implementation Consortium (CPIC) guideline for using CYP2D6 genotyping to guide tamoxifen therapy for breast cancer patients. CYP2D6 metabolizes tamoxifen to its more active metabolite, endoxifen, and patients with
Leonid A Ilchuk et al.
International journal of molecular sciences, 23(22) (2022-11-27)
Inducible Cre-dependent systems are frequently used to produce both conditional knockouts and transgenic mice with regulated expression of the gene of interest. Induction can be achieved by doxycycline-dependent transcription of the wild type gene or OH-tamoxifen-dependent nuclear translocation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service