Skip to Content
Merck

SML2297

Chlorflavonin

≥98% (HPLC), Antibiotic, powder

Synonym(s):

2-(3-Chloro-2-hydroxyphenyl)-5-hydroxy-3,7,8-trimethoxy-4H-1-benzopyran-4-one, F1484

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C18H15ClO7
CAS Number:
Molecular Weight:
378.76
UNSPSC Code:
51111800
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Chlorflavonin, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC(OC)=C(C2=C1C(C(OC)=C(C3=C(O)C(Cl)=CC=C3)O2)=O)OC

InChI

1S/C18H15ClO7/c1-23-11-7-10(20)12-14(22)18(25-3)15(26-17(12)16(11)24-2)8-5-4-6-9(19)13(8)21/h4-7,20-21H,1-3H3

InChI key

JLSQXYITDXJTKL-UHFFFAOYSA-N

Biochem/physiol Actions

Antifungal against certain species of mold fungi and a superior antibiotic than streptomycin against M. tuberculosis by targeting AHAS catalytic subunit IlvB1.
Originally isolated from Aspergillus candidus Link, Chlorflavonin is a non-cytotoxic (up to 100 μM; MRC-5 and THP-1) antibiotic with antifungal activity against certain species of mold fungi (MIC = 0.08 μg/mL against A. amstelodami, A. fumigalus, A. ochraceous; Paecilomyces variotii MIC = 2.5 μg/mL, Botrytis cinerea MIC = 5 μg/mL). Chlorflavonin also exhibits antibiotic activity against M. tuberculosis (MIC90 = 1.56 μM) by targeting acetohydroxyacid synthase (AHAS) catalytic subunit ilvB1. Chlorflavonin is superior to streptomycin against M. tuberculosis inside infected macrophages, synergizes with isoniazid and delamanid, leading to a complete sterilization in liquid cultures.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidja Rehberg et al.
ACS infectious diseases, 4(2), 123-134 (2017-11-08)
The flavonoid natural compound chlorflavonin was isolated from the endophytic fungus Mucor irregularis, which was obtained from the Cameroonian medicinal plant Moringa stenopetala. Chlorflavonin exhibited strong growth inhibitory activity in vitro against Mycobacterium tuberculosis (MIC90 1.56 μM) while exhibiting no
Inge Kjærbølling et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(4), E753-E761 (2018-01-11)
The fungal genus of Aspergillus is highly interesting, containing everything from industrial cell factories, model organisms, and human pathogens. In particular, this group has a prolific production of bioactive secondary metabolites (SMs). In this work, four diverse Aspergillus species (A.
J E Munden et al.
Applied microbiology, 19(5), 718-720 (1970-05-01)
Production of chlorflavonin, a new antifungal antibiotic, by strains of Aspergillus candidus is described. Two wild strains of the fungus had distinctly different chlorflavonin-producing capabilities. One strain produced 25 mug of chlorflavonin per ml per 4 to 5 days in

Global Trade Item Number

SKUGTIN
SML2297-25MG04061835307357
SML2297-5MG04061835307364

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service