Skip to Content
Merck

SML2221

CU-CPT9a

≥98% (HPLC)

Synonym(s):

2-Methyl-4-(7-methoxyl-4-quinolinyl)-phenol, Phenol, 4-(7-methoxy-4-quinolinyl)-2-methyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H15NO2
CAS Number:
Molecular Weight:
265.31
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=CC(C2=CC=NC3=CC(OC)=CC=C23)=CC=C1O

InChI

1S/C17H15NO2/c1-11-9-12(3-6-17(11)19)14-7-8-18-16-10-13(20-2)4-5-15(14)16/h3-10,19H,1-2H3

InChI key

HNYBTVKYLVLWCB-UHFFFAOYSA-N

Biochem/physiol Actions

CU-CPT9a is a potent and selective inhibitor of Toll-like receptor 8 (TLR8). It binds to TLR8 at a site different from current small-molecule TLR7/8 agonists, stabilizing the TLR8 dimer in its resting state, thus preventing TLR8 activation It exhibited an IC50 value of 0.5 nM and blocked TLR8 activation induced by either resiquimod (R848) or ssRNA without affecting the closely related TLR7 or cells expressing other TLRs. CU-CPT9a also suppressed TNF-α levels in a dose-dependent manner in cells from patients with rheumatoid arthritis.
Inhibitor of Toll-like receptor 8 (TLR8)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library