Skip to Content
Merck

SML2162

Telithromycin

≥90% (HPLC), bacterial protein synthesis inhibitor, powder

Synonym(s):

(3aS,4R,7R,9R,10R,11R,13R,15R,15aR)-4-Ethyloctahydro-11-methoxy-3a,7,9,11,13,15-hexamethyl-1-[4-[4-(3-pyridinyl)-1H-imidazol-1-yl]butyl]-10-[[3,4,6-trideoxy-3-dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-2H-oxacyclotetradecino[4,3-d]oxazole-2,6,8,14(1H,7H,9H)-tetrone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H65N5O10
CAS Number:
Molecular Weight:
812.00
NACRES:
NA.77
UNSPSC Code:
12352200
EC Number:
682-750-4
MDL number:
Assay:
≥90% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Telithromycin, ≥90% (HPLC)

assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CCCCN2C=NC(C3=CC=CN=C3)=C2)O[C@]4([C@@]1([H])[C@H](C([C@@H](C[C@]([C@@H]([C@H](C([C@H](C(O[C@@H]4CC)=O)C)=O)C)O[C@H]5[C@@H]([C@H](C[C@H](O5)C)N(C)C)O)(OC)C)C)=O)C)C

InChI

1S/C43H65N5O10/c1-12-33-43(8)37(48(41(53)58-43)19-14-13-18-47-23-31(45-24-47)30-16-15-17-44-22-30)27(4)34(49)25(2)21-42(7,54-11)38(28(5)35(50)29(6)39(52)56-33)57-40-36(51)32(46(9)10)20-26(3)55-40/h15-17,22-29,32-33,36-38,40,51H,12-14,18-21H2,1-11H3/t25-,26-,27+,28+,29-,32+,33-,36-,37-,38-,40+,42-,43-/m1/s1

InChI key

LJVAJPDWBABPEJ-PNUFFHFMSA-N

Biochem/physiol Actions

Ketolide antibiotic
Telithromycin is a ketolide antibiotic, the first to enter clinical use. It is used to treat mild to moderate respiratory infections. Telithromycin binds to the subunit 50S of the bacterial ribosome, interfering with bacterial protein synthesis.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT SE 1

target_organs

Liver,Eyes

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library