Skip to Content
Merck

Skip To

SML1858

AZD2461

≥98% (HPLC)

Synonym(s):

4-[4-Fluoro-3-[(4-methoxypiperidin-1-yl)carbonyl]benzyl]phthalazin-1(2H)-one, 4-[[4-Fluoro-3-[(4-methoxy-1-piperidinyl)carbonyl]phenyl]methyl]-1(2H)-phthalazinone, AZD 2461, AZD-2461

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H22FN3O3
CAS Number:
Molecular Weight:
395.43
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NN=C(CC2=CC=C(F)C(C(N3CCC(OC)CC3)=O)=C2)C4=CC=CC=C41

InChI

1S/C22H22FN3O3/c1-29-15-8-10-26(11-9-15)22(28)18-12-14(6-7-19(18)23)13-20-16-4-2-3-5-17(16)21(27)25-24-20/h2-7,12,15H,8-11,13H2,1H3,(H,25,27)

InChI key

HYNBNUYQTQIHJK-UHFFFAOYSA-N

Biochem/physiol Actions

AZD2461, an olaparib analog, is an orally available, potent and selective PARP1 and PARP2 inhibitor that is a poor substrate for drug transporters. AZD2461 exhibits a high efficacy in olaparib-resistant tumors that overexpress P-glycoprotein.
Orally available, potent and selective PARP1 and PARP2 inhibitor that is a poor substrate for drug transporters

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1674SML0662SML2795
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 30 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters