Skip to Content
Merck

SML1828

EMD-87580

≥98% (HPLC)

Synonym(s):

EMD87580, N-[2-methyl-4,5-bis(methylsulfonyl)-benzoyl]-guanidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H15N3O5S2
CAS Number:
Molecular Weight:
333.38
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

NC(NC(C1=CC(S(=O)(C)=O)=C(S(=O)(C)=O)C=C1C)=O)=N

InChI

1S/C11H15N3O5S2/c1-6-4-8(20(2,16)17)9(21(3,18)19)5-7(6)10(15)14-11(12)13/h4-5H,1-3H3,(H4,12,13,14,15)

InChI key

GROMEQPXDKRRIE-UHFFFAOYSA-N

Application

EMD-87580 has been used as a selective and potent sodium-hydrogen antiporter 1 (NHE-1) inhibitor in cardiac ventricular myocytes to test its effect on the hypertrophic response.

Biochem/physiol Actions

EMD-87580 is a selective inhibitor of the Na+/H+ exchanger subtype 1 (NHE-1), also known as the Na+/H+ antiporter. EMD-87580 has shown cardioprotective effects, and has been investigated as a possible treatment for Duchenne′s muscular dystrophy.
Inhibitor of sodium-hydrogen exchanger-1 (NHE-1)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library