Skip to Content
Merck

Skip To

SML1179

WWL113

≥98% (HPLC)

Synonym(s):

4′-[[[Methyl[[3-(4-pyridinyl)phenyl]methyl]amino]carbonyl]oxy]-[1,1′-Biphenyl]-4-carboxylic acid ethyl ester, Ethyl 4′-((methyl(3-(pyridin-4-yl)benzyl)carbamoyl)oxy)-[1,1′-biphenyl]-4-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C29H26N2O4
CAS Number:
Molecular Weight:
466.53
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCOC(C(C=C1)=CC=C1C(C=C2)=CC=C2OC(N(C)CC3=CC(C4=CC=NC=C4)=CC=C3)=O)=O

InChI

1S/C29H26N2O4/c1-3-34-28(32)25-9-7-22(8-10-25)23-11-13-27(14-12-23)35-29(33)31(2)20-21-5-4-6-26(19-21)24-15-17-30-18-16-24/h4-19H,3,20H2,1-2H3

InChI key

AKIIPHDGVCFVCC-UHFFFAOYSA-N

Biochem/physiol Actions

WWL113 is an inhibitor of mouse Carboxylesterase 3 (Ces3) and human CES1 (orthologue of mCes3), serine hydrolases involved in lipolysis in addition to their activities as liver detoxification enzymes. In a recent study, hCES1 activity was found to be increased two-fold in obese individuals and patients with type 2 diabetes compared to lean subjects, and is thought to generate surplus fatty acids that can deposit ectopically in tissues. WWL113 treatment resulted in major improvement of multiple features of metabolic syndrome and ameliorated obesity-diabetes in mice with lowered levels of nonesterified free fatty acids (NEFAs), triglycerides (TGs), total cholesterol and fasted glucose as well as enhanced glucose tolerance after three weeks of treatment. WWL113 inhibits Ces3 with an IC50 of 120 nM and also the closely related Ces1f with an IC50 of 100 nM. WWL113 inhibits mouse recombinant Ces1, Ces1c, and Abhd6 at 10 μM.
WWL113 is an inhibitor of mouse Carboxylesterase 3 (Ces3) and human CES1 (orthologue of mCes3).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1641SML2874SML1128
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 25 mg/mL, clear

color

white to beige

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters