Skip to Content
Merck

SML1098

Vilazodone hydrochloride

≥98% (HPLC)

Synonym(s):

5-[4-[4-(5-Cyano-1H-indol-3-yl)butyl]-1-piperazinyl]- 2-benzofurancarboxamide hydrochloride, EMD 68843, SB 659746A.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H27N5O2 · HCl
CAS Number:
Molecular Weight:
477.99
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

Cl.N3(CCN(CC3)c4cc5c([o]c(c5)C(=O)N)cc4)CCCCc1c2c([nH]c1)ccc(c2)C#N

InChI

1S/C26H27N5O2.ClH/c27-16-18-4-6-23-22(13-18)19(17-29-23)3-1-2-8-30-9-11-31(12-10-30)21-5-7-24-20(14-21)15-25(33-24)26(28)32;/h4-7,13-15,17,29H,1-3,8-12H2,(H2,28,32);1H

InChI key

RPZBRGFNBNQSOP-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Vilazodone is a serotonin 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SPARI) that is used clinically as an antidepressant for major depressive disorder.
Vilazodone is a serotonin 5-HT1A partial agonist and serotonin-selective reuptake inhibitor (SPARI); antidepressant.


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library