Skip to Content
Merck
All Photos(1)

Documents

SML1084

Sigma-Aldrich

BMS-195614

≥97% (HPLC)

Synonym(s):

4-[5,6-Dihydro-5,5-dimethyl-8-(quinolin-3-yl)naphthalen-2-carboxamido] benzoic acid; 4-[[[5,6-Dhydro-5,5-dimethyl-8-(3-quinolinyl)-2-naphthalenyl]carbonyl]amino]-benzoic acid, BMS 614, BMS-195614, BMS-614, BMS195614, BMS614

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H24N2O3
CAS Number:
Molecular Weight:
448.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1(C)CC=C(C2=CN=C(C=CC=C3)C3=C2)C4=C1C=CC(C(NC5=CC=C(C(O)=O)C=C5)=O)=C4

InChI

1S/C29H24N2O3/c1-29(2)14-13-23(21-15-19-5-3-4-6-26(19)30-17-21)24-16-20(9-12-25(24)29)27(32)31-22-10-7-18(8-11-22)28(33)34/h3-13,15-17H,14H2,1-2H3,(H,31,32)(H,33,34)

InChI key

WGLMBRZXZDAQHP-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-195614 (BMS614) is a potent neutral retinoic acid receptor RARα selective antagonist with an IC50 of 2.5 nM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service