Skip to Content
Merck

SML1053

LMK235

≥98% (HPLC)

Synonym(s):

N-[[6-(Hydroxyamino)-6-oxohexyl]oxy]-3,5-dimethyl-benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H22N2O4
CAS Number:
Molecular Weight:
294.35
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(OCCCCCC(=O)NO)C(=O)c1cc(cc(c1)C)C

InChI

1S/C15H22N2O4/c1-11-8-12(2)10-13(9-11)15(19)17-21-7-5-3-4-6-14(18)16-20/h8-10,20H,3-7H2,1-2H3,(H,16,18)(H,17,19)

InChI key

VRYZCEONIWEUAV-UHFFFAOYSA-N

Biochem/physiol Actions

LMK235 is a histone deacetylase (HDAC) inhibitor with greater potency against HDAC4 and HDAC5 (IC50 = 11.9 and 4.2 nM, respectively) than other HDAC family members (IC50 values = HDAC1 320 nM, HDAC2 881 nM, HDAC6 55.7 nM, and HDAC8 1278 nM). LMK235 potentiates the cytotoxic effects of cisplatin, and sensitizes platinum-drug resistant tumor cell lines to cisplatin toxicity.
LMK235 is a histone deacetylase (HDAC) inhibitor; HDAC4- and HDAC5-preferring.
LMK-235 induces the differentiation of odontoblasts in dental pulp cells. It plays an important role in the regeneration of dental tissue.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library