Skip to Content
Merck
All Photos(1)

Key Documents

SML0972

Sigma-Aldrich

ORM-10103

≥98% (HPLC)

Synonym(s):

2-[(3,4-Dihydro-2-phenyl-2H-1-benzopyran-6-yl)oxy]-5-nitro-pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N2O4
CAS Number:
Molecular Weight:
348.35
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

InChI

1S/C20H16N2O4/c23-22(24)16-7-11-20(21-13-16)25-17-8-10-19-15(12-17)6-9-18(26-19)14-4-2-1-3-5-14/h1-5,7-8,10-13,18H,6,9H2

InChI key

GZONLGPIHCCJOI-UHFFFAOYSA-N

Biochem/physiol Actions

ORM-10103 is a potent, specific inhibitor of the Na(+)/Ca(2+) exchanger, NCX. The compound ORM-10103 inhibits both inward and outward NCX currents with IC50 values of 780 nM, and 960 nM, respectively. ORM-10103 blocks induced arrhythmias in canine cardiac tissue.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service