Skip to Content
Merck
All Photos(1)

Key Documents

SML0954

Sigma-Aldrich

Thiocolchicine

≥97% (HPLC)

Synonym(s):

10-Demethoxy-10-(methylthio)colchicine, 10-Thio-colchicine, N-[(7S)-5,6,7,9-Tetrahydro-1,2,3-trimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-acetamide, NSC 186301

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO5S
CAS Number:
Molecular Weight:
415.50
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

S(C)c1[c](cc2c(cc1)c3c(cc(c(c3OC)OC)OC)CC[C@@H]2NC(=O)C)=O

InChI

1S/C22H25NO5S/c1-12(24)23-16-8-6-13-10-18(26-2)21(27-3)22(28-4)20(13)14-7-9-19(29-5)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1

InChI key

CMEGANPVAXDBPL-INIZCTEOSA-N

Biochem/physiol Actions

Thiocolchicine is an antimitotic alkaloid and apoptosis inducer that inhibits tubulin polymerization and microtubule assembly.
Thiocolchicine is an antimitotic alkaloid and apoptosis inducer.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorota Bartusik et al.
Bioorganic chemistry, 38(1), 1-6 (2009-12-01)
It was shown, that cultured ex vivo human T-Lymphoblastoid (CEM) cells respond to synthesized thiocolchicine and fluorine thiocolchicine derivatives. The preparation of derivatives with substitution at C-3 and C-7 is described. All compounds were used at concentration from 1 nM
R M Chabin et al.
Biochemical and biophysical research communications, 161(2), 544-550 (1989-06-15)
Thiocolchicine, a colchicine analog in which the C-10 methoxy is replaced with a thiomethyl moiety, was shown to bind with high affinity to the colchicine site on tubulin (Ka = 1.07 +/- 0.14 x 10(6) M-1 at 23 degrees C).
A Muzaffar et al.
Journal of medicinal chemistry, 33(2), 567-571 (1990-02-01)
Esterification of the phenolic group in 3-demethylthiocolchicine and exchange of the N-acetyl group with other N-acyl groups or a N-carbalkoxy group afforded many compounds which showed superior activity over the parent drug as inhibitors of tubulin polymerization and of the
R De Vincenzo et al.
Oncology research, 11(3), 145-152 (1999-10-20)
Three new 7-0-substituted deacetamidothiocolchicine derivatives have been evaluated for their antitumor activity against various human tumor cell lines, some of which express the multidrug resistance (MDR) phenotype, for their impact on the cell cycle and their binding to tubulin. Colchicine
R Brecht et al.
Bioorganic & medicinal chemistry, 8(3), 557-562 (2000-03-25)
Several B-ring variations of O-methyl androbiphenyline (8), newly accessible from (-)-(M,7S)-colchicine via photooxygenation and subsequent endoperoxide-transformation, were synthesized and evaluated for their inhibitory effects on tubulin assembly in vitro. The amino-allocolchicinoid (9), a key compound in this study, was transformed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service