Skip to Content
Merck

SML0688

PO1

≥98% (HPLC)

Synonym(s):

2′,3′,6′,7′-Tetrahydro-12′-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-Spiro[isobenzofuran-1(3H)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H32BNO5
CAS Number:
Molecular Weight:
521.41
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

faintly pink to red

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

B8(OC(C(O8)(C)C)(C)C)c1cc2c(cc1)C6(OC(=O)c7c6cccc7)c3c(c4c5c(c3)CCCN5CCC4)O2

InChI

1S/C32H32BNO5/c1-30(2)31(3,4)39-33(38-30)20-13-14-24-26(18-20)36-28-22-11-8-16-34-15-7-9-19(27(22)34)17-25(28)32(24)23-12-6-5-10-21(23)29(35)37-32/h5-6,10,12-14,17-18H,7-9,11,15-16H2,1-4H3

InChI key

KNTNXZHHCPNNGB-UHFFFAOYSA-N

General description

Peroxy orange 1 (PO1) is a H2O2-specific fluorescent probe. PO1 probe is non-trappable and highly permeable in nature.

Application

Peroxy orange 1 (PO1) has been used to detect H2O2 in RtSS (root to shoot silencing) and wild-type Columbia and in rci3-1 and rci3-2 mutants.

Biochem/physiol Actions

PO-1 is a cell-permeable fluorescent H2O2 sensor.
PO-1 is a cell-permeable fluorescent sensor that is highly selective for detecting the presence of H2O2 in living cells.

Legal Information

This product may be used for research purposes only. It is not licensed for resale and may only be used by the buyer. This product may not be used and is not licensed for clinical assays, where the results of such assays are provided as a diagnostic service. If a diagnostic or therapeutic use is anticipated, then a license must be requested from the University of California. The availability of such diagnostic and therapeutic use license(s) cannot be guaranteed from the University of California.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library