Skip to Content
Merck
All Photos(1)

Key Documents

SML0654

Sigma-Aldrich

Entacapone

≥98% (HPLC)

Synonym(s):

2-Cyano-N,N-diethyl-3-(3,4-dihydroxy-5-nitrophenyl)propenamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N3O5
CAS Number:
Molecular Weight:
305.29
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[N+](=O)([O-])c1c(c(cc(c1)\C=C(\C(=O)N(CC)CC)/C#N)O)O

InChI

1S/C14H15N3O5/c1-3-16(4-2)14(20)10(8-15)5-9-6-11(17(21)22)13(19)12(18)7-9/h5-7,18-19H,3-4H2,1-2H3/b10-5+

InChI key

JRURYQJSLYLRLN-BJMVGYQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Entacapone has been used in high-performance bioaffinity chromatography.

Biochem/physiol Actions

Entacapone acts predominantly in the intestine. It increases the bioavailability of levodopa. Levodopa is known to be associated with many complications such as motor fluctuations. COMT inhibitors have been used to overcome these adverse effects of levodopa by reducing its metabolism. Entacapone exhibits its inhibition action on COMT in the peripheral nervous system.
Entacapone is a catechol-O-methyl transferase (COMT) inhibitor. Used in treatment of Parkinson′s disease, entacapone is administered with L-DOPA to inihibit COMT from converting L-DOPA into a compound that cannot cross the blood brain barrier.
Entacapone is a catechol-O-methyl transferase (COMT) inhibitor. Antiparkinson; anti-Dyskinesia.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Principles of Treatment in Parkinson's Disease, 133-133 (2005)
Els F Halff et al.
Molecular imaging, 2022, 4419221-4419221 (2023-02-02)
Positron emission tomography (PET) using the radiotracer [18F]-FDOPA provides a tool for studying brain dopamine synthesis capacity in animals and humans. We have previously standardised a micro-PET methodology in mice by intravenously administering [18F]-FDOPA via jugular vein cannulation and assessment
Alice Petty et al.
Molecular imaging and biology, 25(6), 1054-1062 (2023-10-24)
There is robust evidence that people with schizophrenia show elevated dopamine (DA) synthesis capacity in the striatum. This finding comes from positron emission tomography (PET) studies using radiolabelled l-3,4-dihydroxyphenylalanine (18F-DOPA). DA synthesis capacity also appears to be elevated in the
Entacapone in the treatment of Parkinson's disease
Schrag Anette
Lancet Neurology, 4(6), 366-370 (2005)
Expanding LogP: Present possibilities
Vraka C, et al.
Nuclear Medicine and Biology, 58(6), 20-32 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service