Skip to Content
Merck
All Photos(1)

Documents

SML0530

Sigma-Aldrich

Clonixin

≥98% (HPLC)

Synonym(s):

2-[(3-Chloro-2-methylphenyl)amino]-3-pyridinecarboxylic acid, CBA 93626, Chlonixin, Clonixic acid, Clonixin, Clonixine, NSC 335505, Sch 10304

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11ClN2O2
CAS Number:
Molecular Weight:
262.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

room temp

SMILES string

Cc1c(Cl)cccc1Nc2ncccc2C(O)=O

InChI

1S/C13H11ClN2O2/c1-8-10(14)5-2-6-11(8)16-12-9(13(17)18)4-3-7-15-12/h2-7H,1H3,(H,15,16)(H,17,18)

InChI key

CLOMYZFHNHFSIQ-UHFFFAOYSA-N

Biochem/physiol Actions

Clonixin is a non steroidal anti inflammatory drug (NSAID) with analgesic properties. The compound has been reported to block prostaglandin synthesis, and to block inward calcium currents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Douglas et al.
Journal of food protection, 75(7), 1270-1277 (2012-09-18)
β-Lactam antibiotics are the most commonly used drugs on dairy farms. β-Lactam residues in milk are kept out of the human milk supply with good agricultural practices and mandatory truck screening performed by the dairy industry under Appendix N of
L W Kissell et al.
Journal of dairy science, 95(12), 7151-7157 (2012-10-09)
The objective of this study was to determine if the plasma pharmacokinetics and milk elimination of flunixin (FLU) and 5-hydroxy flunixin (5OH) differ following intramuscular and subcutaneous injection of FLU compared with intravenous injection. Twelve lactating Holstein cows were used
D Goerigk et al.
Tierarztliche Praxis. Ausgabe G, Grosstiere/Nutztiere, 40(2), 112-118 (2012-04-25)
Malignant catarrhal fever (MCF) is a worldwide occurring sporadic disease of cloven-hoofed animals. For the first time, this case report describes clinical and pathological as well as histopathological findings in an alpaca suffering from MCF caused by ovine herpesvirus 2.
David R Paull et al.
Veterinary anaesthesia and analgesia, 39(5), 539-549 (2012-04-25)
To evaluate the analgesic efficacy of the NSAIDs flunixin and meloxicam administered locally to the scrotum before ring castration. Randomised, controlled, prospective study. Forty eight single born male Merino lambs. Lambs, aged approximately 4 weeks, were allocated to four groups
Pharmacokinetic interactions of flunixin meglumine and doxycycline in broiler chickens.
F Yang et al.
Journal of veterinary pharmacology and therapeutics, 36(1), 85-88 (2012-02-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service