Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C14H14O3S
CAS Number:
Molecular Weight:
262.32
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
assay
≥98% (HPLC)
form
powder
color
light orange to dark orange
solubility
DMSO: 10 mg/mL (clear solution)
storage temp.
2-8°C
SMILES string
COCCSC1=C(C)C(=O)c2ccccc2C1=O
InChI
1S/C14H14O3S/c1-9-12(15)10-5-3-4-6-11(10)13(16)14(9)18-8-7-17-2/h3-6H,7-8H2,1-2H3
InChI key
JTCRCMIBIBLUOK-UHFFFAOYSA-N
Biochem/physiol Actions
VK3-OCH3 is an analog of Vitamin K3 (Menadione) with potent antitumor activity. It has been shown to induce G2/M arrest and apoptosis in neuroblastoma cells with much less cytotoxicity towards normal cells. VK3-OCH3 is believed to act through up-regulation of heme oxygenase (HO)-1.
VK3-OCH3 is an analog of Vitamin K3 with potent antitumor activity.
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form powder | form powder | form powder |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| solubility DMSO: 10 mg/mL (clear solution) | solubility DMSO: soluble, chloroform: soluble | solubility DMSO: 10 mg/mL, clear | solubility DMSO: 20 mg/mL, clear |
| color light orange to dark orange | color - | color , faint red to dark red | color , orange to dark orange-red |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




