Skip to Content
Merck
All Photos(1)

Documents

SML0337

Sigma-Aldrich

S-Allyl-L-cysteine

≥98% (HPLC)

Synonym(s):

L-Deoxyalliin, S-allyl-cysteine, S-allylcysteine, SAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2S
CAS Number:
Molecular Weight:
161.22
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -8 to -15°, c = 1 in H2O

color

white to beige

solubility

H2O: >10 mg/mL

storage temp.

−20°C

SMILES string

N[C@@H](CSCC=C)C(O)=O

InChI

1S/C6H11NO2S/c1-2-3-10-4-5(7)6(8)9/h2,5H,1,3-4,7H2,(H,8,9)/t5-/m0/s1

InChI key

ZFAHNWWNDFHPOH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-Allyl-L-cysteine has anti-inflammatory properties. It inhibits prooxidant enzymes and chelates metals. S-Allyl-L-cysteine exhibits hypoglycemic and cholesterol-lowering effects. It prevents apoptosis, lowers the activity of inducible nitric oxide synthase and blocks nuclear factor kappa B activity.

Biochem/physiol Actions

S-allyl-L-cysteine is a sulfur containing amino acid found in garlic with antioxidant, anti-cancer, antihepatotoxic, neuroprotective and neurotrophic activity. S-allyl-L-cysteine has potent activity in several animal models of ischemic injury and Alzhemer′s disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Rojas et al.
The Journal of nutritional biochemistry, 22(10), 937-944 (2010-12-31)
S-Allylcysteine (SAC), the most abundant organosulfur compound in aged garlic extract, has multifunctional activity via different mechanisms and neuroprotective effects that are exerted probably via its antioxidant or free radical scavenger action. The 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine-treated mouse has been the most widely
J Yu et al.
Placenta, 33(6), 487-494 (2012-03-13)
Preeclampsia (PE) is a major cause of fetal growth restriction and perinatal mortality, which involves oxidative stress and vasodilator signaling disorder. S-allyl-L-cysteine (SAC) is one of the most abundant compounds in garlic extracts, and possesses several biological activities. This research
Hayate Javed et al.
Brain research, 1389, 133-142 (2011-03-08)
S-allyl cysteine (SAC), a sulfur containing amino acid derived from garlic, has been reported to have antioxidant, anti-cancer, antihepatotoxic and neurotrophic activity. This study was designed to examine the pre-treatment effects of SAC on cognitive deficits and oxidative damage in
Bioactive Nutraceuticals and Dietary Supplements in Neurological and Brain Disease: Prevention and Therapy (2014)
Man-Hui Pai et al.
The British journal of nutrition, 108(1), 28-38 (2011-10-21)
Oral cancer is prevalent worldwide. Studies have indicated that an increase in the osteopontin (OPN) plasma level is correlated with the progression of oral cancer. Our previous report showed that the aqueous garlic extract S-allylcysteine (SAC) inhibited the epithelial-mesenchymal transition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service