Skip to Content
Merck
All Photos(1)

Documents

SML0184

Sigma-Aldrich

Ginsenoside Rg3

≥98% (HPLC)

Synonym(s):

(3β,12β)-12,20-Dihydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, 20(S)-Ginsenoside-Rg3, Rg3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O13
CAS Number:
Molecular Weight:
785.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +10 to +20°, c = 1 in methanol

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC[C@](C)(O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]3[C@@]4(C)CC[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(C)(C)[C@@H]4CC[C@@]23C

InChI

1S/C42H72O13/c1-21(2)10-9-14-42(8,51)22-11-16-41(7)29(22)23(45)18-27-39(5)15-13-28(38(3,4)26(39)12-17-40(27,41)6)54-37-35(33(49)31(47)25(20-44)53-37)55-36-34(50)32(48)30(46)24(19-43)52-36/h10,22-37,43-51H,9,11-20H2,1-8H3/t22-,23+,24+,25+,26-,27+,28-,29-,30+,31+,32-,33-,34+,35+,36-,37-,39-,40+,41+,42-/m0/s1

InChI key

RWXIFXNRCLMQCD-JBVRGBGGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ginsenoside Rg3 has been used to investigate its modulatory effects on delayed rectifier potassium current (IKr) channels in long QT syndrome (LQTS)2-human induced pluripotent stem cells (hiPSC-CMs).
Ginsenoside Rg3 may be used in cell signaling studies.

Biochem/physiol Actions

Ginsenoside Rg3 is a natural product isolated from Panax ginseng. Similar to other ginsenosides it exhibits cardio protective effects. Ginsenoside Rg3 enhances cardiac, hERG (IKr) and KCNQ (Iks), channel currents by interaction with the channel pore entryway. It also inhibits the palmitate-induced apoptosis of MIN6N8 mouse insulinoma beta-cells through modulating p44/42 MAPK activation. Ginsenoside Rg3 increases the level of the transcription factor Nrf2 and induces the mRNA levels of multidrug resistance-associated protein (Mrp) 1 and Mrp3. Rg3 modulate neuroinflammation by attenuating the activation of microglia in response to systemic LPS treatment. It activates AMPK in HepG2 cells and reduces the lipid accumulation thereby decreasing the risk of cardiovascular disease due to dyslipidemia.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seohyun Lee et al.
International journal of molecular sciences, 13(5), 5729-5739 (2012-07-04)
Cardiovascular disease (CVD) is one of the main causes of mortality worldwide, and dyslipidemia is a major risk factor for CVD. Ginseng has been widely used in the clinic to treat CVD. Ginsenoside Rg3, one of the major active components
Xiaojie Wei et al.
International immunology, 24(7), 465-471 (2012-03-20)
Our previous investigation demonstrated that ginsenoside Rg3 was active in promotion of the immune response. In this study, two epimers, 20(R)-Rg3 and 20(S)-Rg3, were compared for their adjuvant effects on the immune response against ovalbumin (OVA). BALB/c mice were immunized
Jian-Wen Jiang et al.
World journal of gastroenterology, 17(31), 3605-3613 (2011-10-12)
To investigate the anti-tumor function of ginsenoside Rg3 on hepatocellular carcinoma (HCC) in vitro and in vivo, and its mechanism. Hep1-6 and HepG2 cells were treated by Rg3 in different concentrations (0, 50, 100 and 200 μg/mL) in vitro. After
Sun-Min Park et al.
Biological & pharmaceutical bulletin, 35(9), 1546-1552 (2012-09-15)
Neuroinflammation, characterized by activation of microglia and expression of major inflammatory mediators, contributes to neuronal damage in addition to acute and chronic central nervous system (CNS) disease progression. The present study investigated the immune modulatory effects of ginsenoside Rg3, a
Jin-Kwang Kim et al.
Food chemistry, 141(2), 1369-1377 (2013-06-26)
The ginsenoside Rg3(S), which is one of the exceptional components of Korean red ginseng extract, has been known to have anti-cancer, anti-metastatic, and anti-obesity effects. An enzymatic bioconversion method was developed to obtain the ginsenoside Rg3(S) with a high specificity

Articles

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service