Skip to Content
Merck

SML0138

Levalbuterol hydrochloride

≥98% (HPLC)

Synonym(s):

(R)-salbutamol hydrochloride, Levosalbutamol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H21NO3 · HCl
CAS Number:
Molecular Weight:
275.77
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -25 to -35°, c = 0.1 in H2O

storage condition

desiccated

color

white to tan

solubility

H2O: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CC(C)(C)NC[C@H](O)c1ccc(O)c(CO)c1

InChI

1S/C13H21NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,12,14-17H,7-8H2,1-3H3;1H/t12-;/m0./s1

InChI key

OWNWYCOLFIFTLK-YDALLXLXSA-N

Gene Information

human ... ADRB2(154)

Application

Levalbuterol hydrochloride may be used in β2-adrenergic-mediated cell signaling studies.

Biochem/physiol Actions

β-Adrenergic Agonist; Bronchodilator; more active isomer of albutamol
Levalbuterol (levosalbutamol) is the more active isomer of albuterol. Levalbuterol is a β-adrenergic agonist; a bronchodilator used clinically to treat asthma and COPD..
Levalbuterol is (R)-albuterol that exhibits bronchodilator effect and is more effective than racemic albuterol for the treatment of exacerbation of asthma in pediatric populations.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Still not finding the right product?

Explore all of our products under Levalbuterol hydrochloride


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library