Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C24H40N2
CAS Number:
Molecular Weight:
356.59
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
208-897-2
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
assay
≥97% (HPLC)
form
powder
color
white to tan
solubility
ethanol: ≥2 mg/mL
originator
Abbott
storage temp.
2-8°C
SMILES string
C[C@H]1[C@H]2CC[C@H]3[C@@H]4CC=C5C[C@H](CC[C@]5(C)[C@H]4CC[C@]23CN1C)N(C)C
InChI
1S/C24H40N2/c1-16-20-8-9-22-19-7-6-17-14-18(25(3)4)10-12-23(17,2)21(19)11-13-24(20,22)15-26(16)5/h6,16,18-22H,7-15H2,1-5H3/t16-,18-,19+,20+,21-,22-,23-,24-/m0/s1
InChI key
GPLGAQQQNWMVMM-MYAJQUOBSA-N
General description
Conessine is a steroidal alkaloid. It has anti-malarial properties, thus, conessine has been used in treatment of malaria.
Biochem/physiol Actions
Conessine is a plant steroid alkaloid that acts as a potent and specific antagonist of histamine H3 receptors (Ki = 5.37 and 24.5 nM for human and rat receptors, respectively). Conessine is 1860-fold selective for H3 over H4 and does not bind to H1 or H2 receptors. The molecule also binds to the human α2C4 adrenergic receptor (pKi = 7.98).
H3 receptor antagonist
Features and Benefits
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Still not finding the right product?
Explore all of our products under Conessine
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
