Skip to Content
Merck
All Photos(1)

Key Documents

SMB00704

Sigma-Aldrich

Lipopolysaccharides from Proteus mirabilis

purified by phenol extraction

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352211
NACRES:
NA.25

biological source

bacterial (Proteus mirabilis)

Quality Level

form

lyophilized powder

purified by

phenol extraction

impurities

<3.00% protein (Lowry-TCA)

color

white to faint yellow

solubility

water: 0.90-1.10 mg/mL, slightly hazy to hazy, colorless to light yellow

shipped in

ambient

storage temp.

2-8°C

General description

Lipopolysaccharides (LPS′s) are complex glycolipids that are characteristic components of the cell wall of Gram-negative bacteria. They consist of a lipid A moiety linked to an antigenic O-polysaccharide.
Proteus mirabilis is a Gram-negative facultative anaerobic rod-shaped bacterium. Bacteria of the genus Proteus of the family Enterobacteriaceae are opportunistic human pathogens responsible for wound and burn infections as well as skin, eye, ear, nose, throat, urinary tract, and gastrointestinal infections and bacteremias.

Application


  • Lipopolysaccharide structure modulates cationic biocide susceptibility and crystalline biofilm formation in Proteus mirabilis.: The study explores how variations in lipopolysaccharide structure affect the susceptibility of Proteus mirabilis to biocides and its ability to form crystalline biofilms, informing strategies to combat biofilm-associated infections (Clarke et al., 2023).

Biochem/physiol Actions

Lipopolysaccharide (LPS) and its lipid A moiety stimulate cells of the innate immune system by the toll-like receptor 4 (TLR4), a member of the toll-like receptor protein family, which recognizes common pathogen-associated molecular patterns (PAMPs). Lipid A plays a role in the biological properties attributed to endotoxic LPS. LPS is a highly immunogenic antigen that has the ability to enhance immune responses.The outer-membrane lipopolysaccharide is considered an important virulence factor of Proteus. It has been suggested that the immunological response against P. mirabilis, LPS might play a role in rheumatoid arthritis. A possible correlation between the abundance of P. mirabilis in the intestine and obesity was suggested recently. The purified LPS from P. mirabilis OXK is used in the Weil-Felix test for the diagnosis of rickettsiosis (scrub typhus) caused by the bacterium Orientia (Rickettsia) tsutsugamushi.

Preparation Note

Soluble in water and cell culture medium DMEM (1 mg/mL) yielding a clear to slightly hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Distribution of core oligosaccharide types in lipopolysaccharides from Escherichia coli.
Amor K, et al.
Infection and Immunity, 68(3), 1116-1124 (2000)
Analysis of Lipopolysaccharides of Gram-Negative Bacteria
Mayer H, Tharanathan RN, Weckesser J
Methods in Microbiology, 18, 157-207 (1985)
A S Swierzko et al.
Biochemistry. Biokhimiia, 62(1), 21-27 (1997-01-01)
Sugar analysis and 1H- and 13C-NMR spectroscopic studies showed that various strains of Proteus mirabilis OXK used as antigens in the Weil-Felix test for serodiagnosis of rickettsiosis (scrub typhus) produce lipopolysaccharides (LPSs) with the same O-specific polysaccharide chain having the
Molecular basis for structural diversity in the core regions of the lipopolysaccharides of Escherichia coli and Salmonella enterica.
Heinrichs D E, et al.
Molecular Microbiology, 30(2), 221-232 (1998)
Novel adjuvants & delivery vehicles for vaccines development: a road ahead.
Mohan T, et al.
The Indian Journal of Medical Research, 138(5), 779-795 (2013)

Articles

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Explore the structure, function, and diverse applications of Lipopolysaccharides. Discover their role in bacteria, serological specificity, and research potential.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service