Skip to Content
Merck
All Photos(1)

Documents

SMB00607

Sigma-Aldrich

Amoxicillin trihydrate: potassium clavulanate (4:1)

Synonym(s):

Amoxicillin Trihydrate: Potassium Clavulanate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51285005
NACRES:
NA.85

form

powder

Quality Level

storage condition

(Keep container tightly closed in a dry and well-ventilated place. Never allow produc to get in contact with water during storage.)

color

white to yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

General description

Amoxicillin trihydrate is a semisynthetic antibiotic, that is used as a medicine.

Application

Amoxicillin trihydrate: potassium clavulanate (4:1) has been used to study its effect on the viral déjà vu disease (neonatal intracerebral (i.c.) infection) in mice model.

Biochem/physiol Actions

Amoxicillin trihydrate is a broad-spectrum, β-lactam antibiotic. It inhibits the cross-linkage between linear peptidoglycan polymer chains that are the major component of both Gram-positive and Gram-negative bacteria. Since amoxicillin is degraded by β-lactamases, it does not show activity against bacteria which produce these enzymes.

Clavulanate is a β-lactam antibiotic related to the penicillins. Clavulanate competitively and irreversibly inhibits a wide variety of β-lactamases found in bacteria that are resistant to penicillins and cephalosporins.

Formulations of amoxicillin with clavulanic acid prevent the degradation of amoxicillin by β-lactamase enzymes. This increases amoxicillin′s antibacterial activity against bacteria normally resistant to β-lactam antibiotics.

Packaging

1G

Components

Material is a powdered mixture of 4 parts amoxicillin trihydrate to 1 part potassium clavulanate.

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place. Never allow produc to get in contact with water during storage.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Never allow product to get in contact with water during storage.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Resp. Sens. 1 - Self-heat. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neurons under T Cell Attack Coordinate Phagocyte-Mediated Synaptic Stripping
Di Liberto G, et al.
Cell (2018)
Preparation and characterization of Sn/Zn/TiO2 photocatalyst for enhanced amoxicillin trihydrate degradation
Mohammadi R, et al.
Desalination and Water Treatment, 53(7), 1995-2004 (2015)
R N Brogden et al.
Drugs, 18(3), 169-184 (1979-09-01)
Amoxycillin, an acid stable semisynthetic penicillin shown to be effective against a wide range of infections when given orally, is now available for intramuscular and intravenous injection. Amoxycillin has an antibacterial spectrum and level of activity essentially the same as
In vitro antimicrobial activity of 6(D(-) -amino-p-hydroxyphenylacetamido) penicillanic acid, a new semisynthetic penicillin.
H C Neu et al.
Antimicrobial agents and chemotherapy, 10, 407-410 (1970-01-01)
A P Ball et al.
Lancet (London, England), 1(8169), 620-623 (1980-03-22)
Clavulanic acid is an inhibitor of certain beta-lactamases, notably those enzymes which inactivate the penicillins and cause resistance of important bacteria to these antibiotics. Microbiological, pharmacological, and clinical studies show that clavulanic acid in combination with amoxycillin is effective and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service