Skip to Content
Merck
All Photos(1)

Key Documents

SMB00118

Sigma-Aldrich

8-Acetylharpagide

≥85% (LC/MS-ELSD)

Synonym(s):

Harpagide 8-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O11
CAS Number:
Molecular Weight:
406.38
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥85% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(=O)O[C@@]1(C)C[C@@H](O)[C@]2(O)C=CO[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)[C@H]12

InChI

1S/C17H26O11/c1-7(19)28-16(2)5-9(20)17(24)3-4-25-15(13(16)17)27-14-12(23)11(22)10(21)8(6-18)26-14/h3-4,8-15,18,20-24H,5-6H2,1-2H3/t8-,9-,10-,11+,12-,13-,14+,15+,16+,17-/m1/s1

InChI key

CAFTUQNGDROXEZ-XBDCZORHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-hong Zhang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 27(3), 206-208 (2003-05-31)
To investigate the chemical constituents of Galeopsis bifida. Various chromatographic techniques were employed for isolation and purification of the constituents. The structures were elucidated by chemical and spectral analyses. Ten compounds were isolated and identified as chrysoeriol(1), 5,7-dihydroxy-3',4'-dimethoxyflavone(2), hederagenin(3), daucosterol(4)
Dang-Kun Yue et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(4), 289-290 (2006-05-19)
To establish a method of the quantitative determination of acetylharpagide in Ajuga decumbens. The chromatographic conditions were as follows: a Phenomenex Luna C18 column was used, the mobile phase was composed of acetontrile-water (15:85), the flow rate was 1.0 mL
A Elbrecht et al.
Insect biochemistry and molecular biology, 26(6), 519-523 (1996-06-01)
We have identified a novel nonsteroidal ecdysteroid agonist. This compound was isolated from a methanol extract of Ajuga reptans L. (Lamiaceae) and the structure was identified by spectroscopic methods as 8-O-acetylharpagide. We have characterised this compound as an ecdysteroid agonist
Binyu Wen et al.
Journal of ethnopharmacology, 147(2), 503-508 (2013-04-03)
Ajuga decumbens Thunb is a medicinal plant native to China popularly used to treat chronic pelvic inflammation and hysteromyoma. Its main bioactive components are iridoid glycosides, such as 8-O-acetylharpagide and harpagide that had presented antibacterial, anti-inflammatory, and antiviral activities. To
E Abdel-Sattar
Archives of pharmacal research, 21(6), 785-786 (1998-12-30)
The aerial parts of Teucrium yemense yielded two iridoid glycosides. Their structures were elucidated by spectral means as teucardosid and 8-O-acetylharpagid.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service