SMB00090
Vulgarin
≥90% (LC/MS-ELSD)
Synonym(s):
Barrelin, Judaicin, Judaicin (eudesmane naphthofuran), Tauremisin, Tauremizin, Vulgarin
About This Item
Assay
≥90% (LC/MS-ELSD)
form
solid
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
−20°C
SMILES string
C[C@H]1[C@@H]2CC[C@]3(C)[C@@H]([C@H]2OC1=O)[C@](C)(O)C=CC3=O
InChI
1S/C15H20O4/c1-8-9-4-6-14(2)10(16)5-7-15(3,18)12(14)11(9)19-13(8)17/h5,7-9,11-12,18H,4,6H2,1-3H3/t8-,9-,11-,12+,14-,15+/m0/s1
InChI key
NGPDZEACIWDCKX-WUDKWMPASA-N
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service