Skip to Content
Merck
All Photos(1)

Key Documents

SMB00019

Sigma-Aldrich

N-Acetyltyramine

≥95% (LC/MS-ELSD)

Synonym(s):

N-(2-(4-Hydroxyphenyl)ethyl)acetamide, N-(p-Hydroxyphenethyl)acetamide, N-Acetyl tyramine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO2
CAS Number:
Molecular Weight:
179.22
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

N(CCc1ccc(cc1)O)C(=O)C

InChI

1S/C10H13NO2/c1-8(12)11-7-6-9-2-4-10(13)5-3-9/h2-5,13H,6-7H2,1H3,(H,11,12)

InChI key

ATDWJOOPFDQZNK-UHFFFAOYSA-N

General description

Natural product derived from fungal source.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W S Garcez et al.
Journal of agricultural and food chemistry, 48(8), 3662-3665 (2000-08-24)
Two saprophytic fungi (Mucor ramosissimus and Rhizopus sp.) were tested for their ability to induce phytoalexin production by seeds of frog-eye leaf spot and stem canker-resistant and -susceptible soybean (Glycine max L.) cultivars. Only M. ramosissimus was shown to elicit
P H Yu et al.
Canadian journal of biochemistry, 57(10), 1204-1209 (1979-10-01)
The N-acylation of tyramine isomers and other biogenic amines has been studied. The liver exhibits the highest activity towards tyramines, while the brain exhibits a low but significant activity. In the brain, tyramine N-acylation activity was heterogenously distributed. The arylamine
S Friström et al.
Acta pharmacologica et toxicologica, 40(2), 247-258 (1977-02-01)
The effect of five sympathomimetic amines and some of their acetyl derivatives on the blood pressure of the rat was determined on the left carotid artery. After pretreatment with chlorisondamine (1 mg/kg subcutaneously) the blood pressure rise by sympathomimetic amines
A K Mir et al.
Journal of neurochemistry, 36(2), 441-446 (1981-02-01)
N-Acetyltyramine, N-acetyldopamine and N-acetyloctopamine were the major products when either L-[3H]tyrosine or [3H]tyramine were incubated with thoracic ganglia of the desert locust, Schistocerca gregaria. No label was incorporated into L-DOPA under these conditions, although 2-3% of the radioactivity could be
R G Macfarlane et al.
Journal of chromatography, 532(1), 13-25 (1990-10-26)
The N-acetylated metabolites of p-tyramine, p-octopamine and dopamine were identified unambiguously and quantitatively determined in a single ventral thoracic nerve cord of the locust, Schistocerca gregaria, by gas chromatography-negative-ion chemical ionisation mass spectrometry (GC-NICIMS). Deuterium-labelled analogues of each compound were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service