Skip to Content
Merck

SIC002

MISSION® siRNA Universal Negative Control #2

Synonym(s):

MISSION® siRNA Universal Negative Control

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

storage temp.

−20°C

Application

Negative controls are an essential part of every siRNA experiment, and here we offer two sizes for your convenience. This negative control has been tested in vitro in Human, Mouse, and Rat cells.

Legal Information

All siRNA oligos are sold under license from the Massachusetts Institute of Technology.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library