Skip to Content
Merck
All Photos(1)

Documents

SHC016

Sigma-Aldrich

MISSION® pLKO.1-puro Non-Target shRNA Control Plasmid DNA

Targets no known genes from any species

Synonym(s):

MISSION® Control Vectors, negative control, negative shRNA control, non-target control, non-target shRNA, non-target shRNA control, shRNA control

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106609
NACRES:
NA.51

product line

MISSION®

concentration

500 ng/μL in TE buffer; DNA (10μg of plasmid DNA)

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The MISSION pLKO.1-puro Non-Target shRNA Control Plasmid DNA is a lentivirus plasmid vector. The vector contains an shRNA insert that does not target any known genes from any species, making it useful as a negative control in experiments using the MISSION shRNA library clones. This allows one to examine the effect of transfection of a short-hairpin on gene expression and interpret the knockdown effect seen with shRNA clones. Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells respectively. In addition, self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids. The Non-Target shRNA Control Plasmid DNA is provided as 10 μg of plasmid DNA in Tris-EDTA (TE) buffer at a concentration of 500 ng/μl.

Application

MISSION® pLKO.1-puro non-target shRNA control plasmid DNA has been used as a control during transduction:
  • in tumor cells for multicolour imaging
  • in human adult low calcium temperature keratinocytes
  • in mouse embryonic fibroblasts, to study the biological functions of IP6K1 (inositol hexakisphosphate kinase)
  • to study the function of Zac1 (zinc finger protein regulating apoptosis and cell cycle arrest) expression in astroglial differentiation
To see more application data, protocols, vector maps visit sigma.com/shrna.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion of inositol hexakisphosphate kinase 1 (IP6K1) reduces cell migration and invasion, conferring protection from aerodigestive tract carcinoma in mice
Jadav RS, et al.
Cellular Signalling, 28(8), 1124-1136 (2016)
Peroxiredoxin 2 nuclear levels are regulated by circadian clock synchronization in human keratinocytes
Avitabile D, et al.
The International Journal of Biochemistry & Cell Biology, 53(7580), 24-34 (2014)
Zac1 regulates astroglial differentiation of neural stem cells through Socs3
Schmidt EU, et al.
Stem Cells, 31(8), 1621-1632 (2013)
Sophie Weil et al.
Neuro-oncology, 19(10), 1316-1326 (2017-04-19)
Primary and adaptive resistance against chemo- and radiotherapy and local recurrence after surgery limit the benefits from these standard treatments in glioma patients. Recently we found that glioma cells can extend ultra-long membrane protrusions, "tumor microtubes" (TMs), for brain invasion
Brain tumour cells interconnect to a functional and resistant network
Osswald M, et al.
Nature, 528(7580), 93-93 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service