Skip to Content
Merck
All Photos(1)

Key Documents

SCP0084

Sigma-Aldrich

Caspase 3 Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H39N5O12
Molecular Weight:
673.67
UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

Caspase 3 Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Asp-Glu-Val-Asp-4M2NA

General description

Caspase 3 belongs to the family of cysteinyl proteases, which regulate apoptosis. It is encoded by CASP-3 gene. Caspase 3 is important for normal brain development. Caspase 3 is involved in growth stimulation of surviving tumor cells. It is associated with neuronal apoptosis in Alzheimer′s disease.

Application

Caspase 3 Substrate has been used to determine caspase activity in esophageal squamous cell carcinoma EC109 cell line.
Caspase 3, a cysteine protease activator of caspase 6 and caspase 7, metabolizes amyloid-β 4A precursor protein. Caspase 3 may be detected using a variety of chromogenic and fluorogenic peptide substrates built around the DxxD (asp-x-x-asp) sequences DMQD and DEVD such as Ac-DMQD-pNa (Acetyl-Asp-Met-Gln-Asp-p- nitroanalide), chromogenic; Ac-DMQD-AMC (Acetyl-Asp-Met-Gln-Asp- amino-4-methylcoumarin), fluorogenic and Ac-DEVD-4M2Na.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caspase-3-generated fragment of gelsolin: effector of morphological change in apoptosis
Kothakota S, et al.
Science, 278(5336), 294-298 (1997)
Caspase 3-mediated stimulation of tumor cell repopulation during cancer radiotherapy
Huang Q, et al.
Nature Medicine, 17(7), 860-860 (2011)
Resveratrol-induced apoptosis is enhanced by inhibition of autophagy in esophageal squamous cell carcinoma
Tang Q, et al.
Cancer Letters, 336(2), 325-337 (2013)
Caspase-3 is enriched in postsynaptic densities and increased in Alzheimer's disease
Louneva N, et al.
The American Journal of Pathology, 173(5), 1488-1495 (2008)
Emerging roles of caspase-3 in apoptosis
Porter AG and Janicke RU
Cell Death and Differentiation, 6(2), 99-99 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service