SCJ4600031
CF™ 594 succinimidyl ester
suitable for fluorescence, ≥90% (TLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
Assay
≥90% (TLC)
form
solid
mol wt
~729 g/mol (free acid)
solubility
DMF: soluble
DMSO: soluble
H2O: soluble
ε (extinction coefficient)
115000 at 593 nm
absorbance ratio
A280/593 nm 0.08
fluorescence
λex 532 or 561 nm (Flow cytometry laser line)
λex 593 nm; λem 614 nm
λex 594 nm (Microscopy laser line)
suitability
suitable for fluorescence
shipped in
wet ice
storage temp.
−20°C
General description
CF™594 is a deep red fluorescent dye spectrally similar to Alexa Fluor™ 594 and Texas Red® dye. When conjugated to proteins, CF™594 is significantly brighter than Alexa Fluor™ 594 due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability, making it ideal for demanding applications such as confocal microscopy and single molecule imaging. These properties make CF™594 the best deep-red dye for labeling proteins and nucleic acids. The dye is particularly useful in combination with our blue fluorescent CF™350, green fluorescent CF™488A and far red CF™647 for multi-color imaging.
Features and Benefits
1. Yields the brightest antibody conjugates among spectrally similar dyes; 2. Extremely photostable; 3. Highly water-soluble
Preparation Note
Protect from light and moisture
Reconstitution
Dissolve in DMSO
Legal Information
This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Texas Red is a registered trademark of Life Technologies
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service