Skip to Content

SCA20

Sucrose Assay Kit

sufficient for 20 assays

Synonym(s):

Sucrose Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12164500
EC Number:
200-334-9
NACRES:
NA.84
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 20 assays

Quality Segment

analyte chemical class(es)

sugars (sucrose)

technique(s)

photometry: suitable

application(s)

cannabis testing
food and beverages
general analytical

storage temp.

2-8°C

General description

Sucrose is a disaccharide, naturally occurring carbohydrate found in plants and a major product during photosynthesis.

Application

This kit is for the quantitative, enzymatic determination of sucrose in food and other materials. Sucrose is hydrolyzed to glucose and fructose by invertase. Glucose and fructose are phosphorylated by adenosine triphosphate (ATP) in the reaction catalyzed by hexokinase. Glucose-6-phosphate (G6P) is then oxidized to 6-phosphogluconate in the presence of nicotinamide adenine dinucleotide (NAD) in a reaction catalyzed by glucose-6-phosphate dehydrogenase (G6PDH). During this oxidation, an equimolar amount of NAD is reduced to NADH. The consequent increase in absorbance at 340 nm is directly proportional to sucrose concentration.


Kit Components Only

Product No.
Description

  • glucose assay reagent 50 mL

  • sucrose assay reagent 1 mL/vial

  • sucrose standard 500 mg

Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library