Skip to Content
Merck

SAE0214

Leucoagglutinin from P. vulgaris, recombinant

Expressed in P. pastoris, lyophilized powder, suitable for cell culture

Synonym(s):

PHA-L, Phytohemagglutinin-L, Red Kidney Bean Lectin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.21
UNSPSC Code:
12352202
Form:
lyophilized powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

lyophilized powder

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

General description

Lectin from Red Kidney Bean has long been used in a wide variety of applications that require carbohydrate binding including but not limited to T-cell proliferation, leucoagglutination, glycoconjugate capture and more. The native form of lectin from Red Kidney Bean is composed of 2 isolectins (PHA-L and PHA-E) which combine in heterotetromeric combinations that elicit a variety of properties. This recombinant form is strictly the leucoagglutinin (PHA-L) isolectin only, so the erythroagglutinin (PHA-E) isolectin is absent in this product. This allows for more selective performance because there is no chance of PHA-E contamination as seen in native source products.

Biochem/physiol Actions

Kidney bean lectin has a history of being used as a phytohemagglutinin because of its ability to agglutinate blood cells. This isolectin (PHA-L) specficially is the Leucoagglutinin type and can be used for leucoagglutination and as a mitogen. As a lectin, it is known to have some selectivity for binding of β1-6 branched N-glycans.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library