Skip to Content
Merck
All Photos(1)

Key Documents

SAE0050

Sigma-Aldrich

Laccase from Aspergillus sp.

Synonym(s):

Laccase from Aspergillus sp., Novozym 51003

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]1(c(c(cc1C)C=O)C)CC

InChI

1S/C9H13NO/c1-4-10-7(2)5-9(6-11)8(10)3/h5-6H,4H2,1-3H3

InChI key

NWDZDFOKSUDVJV-UHFFFAOYSA-N

General description

Laccase EC 1.10.3.2, a glycoprotein, is an extracellular multicopper enzyme and is considered as a metal.[1][2] Laccase is widely distributed in fungi and also found among the higher plants, bacteria and insects.[1][2]

Biochem/physiol Actions

Laccase oxidizes aromatic and nonaromatic compounds. Various compounds are used for detecting laccase production. These include guaiacol, syringaldazine and polymeric dyes like remazol brilliant blue-R. Laccase is involved in lignin degradation and thereby has industrial as well as food applications. Laccase is commonly used for delignification, dye bleaching, paper processing, waste detoxification, textile dye transformation, plant fiber modification and ethanol production.[3]

Unit Definition

One Unit: LAMU (Laccase Unit). 1 LAMU is defined as the amount of enzyme which oxidizes 1 micromole of syringaldazine per minute, at pH 7.5 and 30 deg C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening and induction of laccase activity in fungal species and its application in dye decolorization.
Kumar VV
African Journal of Microbiology Research, 5, 1261-1267 (2011)
Microwave-assisted covalent immobilization of enzymes on inorganic surfaces.
Plagemann, R., et al.
Engineering in Life Sciences, 14, 493-499 (2014)
Laccase: new functions for an old enzyme.
Mayer A M and Staples R C
Phytochemistry, 60(6), 551-565 (2002)
Screening and induction of laccase activity in fungal species and its application in dye decolorization.
Kumar V V, et al.
African Journal of Microbiology Research, 5(11), 1261-1267 (2011)
Faustine Lorquin et al.
Scientific reports, 11(1), 8538-8538 (2021-04-22)
Pyomelanin is a polymer of homogentisic acid synthesized by microorganisms. This work aimed to develop a production process and evaluate the quality of the pigment. Three procedures have been elaborated and optimized, (1) an HGA-Mn2+ chemical autoxidation (PyoCHEM yield 0.317 g/g

Questions

1–2 of 2 Questions  
  1. Wie hoch ist die Konzentration der Enzyme in der Lösung (z. B. in ug/ml)?

    1 answer
    1. The concentration (and density) of laccase is 1.05 g/ml. The composition as follows:
      Laccase 3%
      Propylene glycol 25%
      Sucrose/Glucose 4%
      Glycine 2%
      Water 66%

      Helpful?

  2. In che solvente è diluito il prodotto?

    1 answer
    1. This product is prepared in water containing approximately 25% propylene glycol, 4% D-glucose/Sucrose and 2% Glycine.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service