Skip to Content
Merck
All Photos(1)

Documents

SAB5300407

Sigma-Aldrich

Monoclonal Anti-OLIG2 antibody produced in mouse

clone 1G11, ascites fluid

Synonym(s):

BHLHB1, OLIGO2, PRKCBP2, RACK17, bHLHe19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1G11, monoclonal

mol wt

32 kDa

species reactivity

human

technique(s)

direct ELISA: 1:10,000
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OLIG2(10215)

Immunogen

Purified recombinant fragment of human OLIG2 expressed in E.coli.
Mouse monoclonal antibody raised against OLIG2

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianghong Man et al.
Cell stem cell, 22(1), 104-118 (2017-12-05)
Tumor hypoxia is associated with poor patient survival and is a characteristic of glioblastoma. Notch signaling is implicated in maintaining glioma stem-like cells (GSCs) within the hypoxic niche, although the molecular mechanisms linking hypoxia to Notch activation have not been
Jie Qu et al.
Neural regeneration research, 18(8), 1750-1756 (2023-02-09)
Piezo1 is a mechanically-gated calcium channel. Recent studies have shown that Piezo1, a mechanically-gated calcium channel, can attenuate both psychosine- and lipopolysaccharide-induced demyelination. Because oligodendrocyte damage and demyelination occur in intracerebral hemorrhage, in this study, we investigated the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service