Skip to Content
Merck
All Photos(4)

Key Documents

SAB5200047

Sigma-Aldrich

Monoclonal Anti-SHANK3 antibody produced in mouse

clone S72-16, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-Prosap2, Anti-SHANK3, S69-46, Anti-Shank3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S72-16, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 190 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

Detects ~190 kDa. No cross-reactivity against Shank1 or Shank2.

Immunogen

Synthetic peptide amino acids 840-857 of rat Shank3, Accession Number Q9JLU4

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meghan Kerrisk Campbell et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(23), 5289-5301 (2018-05-08)
Mutations or altered protein levels of SHANK3 are implicated in neurodevelopmental disorders such as Phelan-McDermid syndrome, autism spectrum disorders, and schizophrenia (Guilmatre et al., 2014). Loss of SHANK3 in mouse models results in decreased synapse density and reduction in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service