Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200017

Sigma-Aldrich

Monoclonal Anti-HIF-1 α antibody produced in mouse

clone ESEE122, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-ARNT interacting protein, Anti-HIF1 alpha, ESEE122, Anti-HIF1A, Anti-Hypoxia inducible factor 1 alpha, Anti-MOP1, Anti-PASD8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ESEE122, monoclonal

form

buffered aqueous glycerol solution

species reactivity

bovine, mouse, human, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Hif1a(15251)

General description

Hypoxia-inducible factor-1 α (HIF1 α) gene encodes the α subunit of HIF-1. HIF-1α contains oxygen-dependent degradation (ODD) domain that mediates the interaction between HIF-1α and the E3 ubiquitin ligase complex, which in turn facilitates continuous poly ubiquitination of HIF-1α in normoxic cells. The gene encoding it is localized on human chromosome 14q23.2. HIF1 α belongs to the basic helix-loop-helix family of transcription factors.

Specificity

Specific for HIF1-alpha.

Immunogen

Recombinant fragment corresponding to amino acids 329-530

Application

Monoclonal Anti-HIF-1 α antibody produced in mouse has been used in siRNA assay and western blot analysis.

Biochem/physiol Actions

Hypoxia-inducible factor-1(HIF1) plays an essential role in cellular and systemic homeostatic responses to hypoxia. During hypoxia, the two subunits of this factor undergo post-translational modifications which in turn promote transactivation. HIF-1α gene loss is associated with development of nonpapillary renal carcinomas. The encoded protein functions as a key regulator of the vascular endothelial growth factor (VEGF) under hypoxic response in human stromal cells in vitro. HIF-1α protein expression is often seen in invasive breast cancer. Experimental data demonstrates that downregulation of HIF-1α decreases tumorigenicity of MCF-7 (human breast adenocarcinoma cell line) cells and propose a promising combination of both anti-HIF-1 strategy and traditional chemotherapy to improve cancer treatment.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knockdown of hypoxia-inducible factor-1alpha in breast carcinoma MCF-7 cells results in reduced tumor growth and increased sensitivity to methotrexate.
Li J
Biochemical and Biophysical Research Communications, 342, 1341-1351 (2006)
Multiple gastrointestinal stromal and other tumors caused by platelet-derived growth factor receptor alpha gene mutations: a case associated with a germline V561D defect.
Pasini B
The Journal of Clinical Endocrinology and Metabolism, 92, 3728-3732 (2007)
Regulation of hypoxia-inducible factor 1alpha expression and function by the mammalian target of rapamycin.
Hudson CC
Molecular and Cellular Biology, 22, 7004-7014 (2002)
Propofol Through Upregulating Caveolin-3 Attenuates Post-Hypoxic Mitochondrial Damage and Cell Death in H9C2 Cardiomyocytes During Hyperglycemia.
Deng F
Cellular Physiology and Biochemistry, 44, 279-292 (2017)
Yang Yang et al.
Cell death & disease, 13(6), 533-533 (2022-06-08)
Benign prostatic hyperplasia (BPH) is the most common and progressive urological disease in elderly men worldwide. Epidemiological studies have suggested that the speed of disease progression varies among individuals, while the pathophysiological mechanisms of accelerated clinical progression in some BPH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service