Skip to Content
Merck
All Photos(6)

Documents

SAB4504721

Sigma-Aldrich

Anti-phospho-p47 phox (pSer345) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CGD1, Anti-NCF1A, Anti-NOXO2, Anti-SH3PXD1A, Anti-p47phox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 44 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer345)

Gene Information

human ... NCF1(653361)

General description

The neutrophil cytosolic factor 1 (NCF1), also known as p47phox gene, spanning 15kb with 11 exons, is mapped to human chromosome 7q11.23. The gene codes for a 390 amino acid protein, which is a novel component of the NADPH oxidase complex.

Immunogen

The antiserum was produced against synthesized peptide derived from human p47 phox around the phosphorylation site of Ser345.

Immunogen Range: 311-360

Biochem/physiol Actions

Neutrophil cytosolic factor 1 (NCF1) stimulates the production of reactive oxygen species (ROS) under hyperoxic condition. The above process can be inhibited by the NCF1 inhibitor diphenylene iodonium (DPI) and apocynin. Mutation in the gene leads to a rare congenital immune deficiency called, chronic granulomatous disease (CGD). Deletion of NCF1 gene, which is a vital component of NADPH oxidase complex implicated in generation of oxidative stress, leads to Williams syndrome. It also act as a modifier of vascular stiffness.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beth A Kozel et al.
Hypertension (Dallas, Tex. : 1979), 63(1), 74-79 (2013-10-16)
Williams syndrome is caused by the deletion of 26 to 28 genes, including elastin, on human chromosome 7. Elastin insufficiency leads to the cardiovascular hallmarks of this condition, namely focal stenosis and hypertension. Extrapolation from the Eln(+/-) mouse suggests that
Astghik Hayrapetyan et al.
Biochimica et biophysica acta, 1832(10), 1662-1672 (2013-05-22)
Chronic granulomatous disease (CGD) is a rare congenital disorder in which phagocytes cannot generate superoxide (O2(-)) and other microbicidal oxidants due to mutations in one of the five components of the O2(-)-generating NADPH oxidase complex. The most common autosomal subtype
Lingping Zhang et al.
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology, 32(1), 59-62 (2016-01-06)
To observe the effects of NADPH oxidase inhibitor diphenylene iodonium (DPI) and apocynin on the generation of reactive oxygen species (ROS) induced by p47phox and the mechanism of p47phox-induced ROS production under hyperoxic conditions. Peripheral blood mononuclear cells (PBMCs) were
U Francke et al.
American journal of human genetics, 47(3), 483-492 (1990-09-01)
Chronic granulomatous disease (CGD) is a heterogeneous group of inherited disorders of impaired superoxide production in phagocytes. The most common X-linked recessive form involves the CYBB locus in band Xp21.1 that encodes the membrane-bound beta subunit of the cytochrome b558
Duo-Yao Cao et al.
Science translational medicine, 13(604) (2021-07-30)
Angiotensin-converting enzyme inhibitors (ACEIs) are used by millions of patients to treat hypertension, diabetic kidney disease, and heart failure. However, these patients are often at increased risk of infection. To evaluate the impact of ACEIs on immune responses to infection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service