Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503891

Sigma-Aldrich

Anti-phospho-Ku80 (pThr714) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 82 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... XRCC5(7520)

Immunogen

The antiserum was produced against synthesized peptide derived from human Ku80 around the phosphorylation site of Thr714.

Immunogen Range: 683-732

Biochem/physiol Actions

The gene XRCC5 (X-ray repair cross complementing 5) is mapped to human chromosome 2q35. It is part of the non-homologous end joining double-strand break repair pathway. XRCC5 helps in damage recognition. Mutations in the gene are associated with risk of glioma and chronic obstructive pulmonary disease (COPD). Mutation in it might also be linked with gastric cancer development, particularly in cases of family history. XRCC5 is also involved in alcohol dependence. In addition, it is a part of the OCFRE (osteocalcin (OC) fibroblast growth factor (FGF) response element (FRE))-binding complex, thereby controlling the gene expression of osteocalcin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regional assignment of a human DNA repair gene (XRCC5) to 2q35 by X-ray hybrid mapping.
Chen DJ, et al.
Genomics, 21, 423-427 (1994)
XRCC5 as a risk gene for alcohol dependence: evidence from a genome-wide gene-set-based analysis and follow-up studies in Drosophila and humans.
Juraeva D, et al.
Neuropsychopharmacology, 40, 361-371 (2015)
Regulation of osteocalcin gene expression by a novel Ku antigen transcription factor complex.
Willis DM, et al.
The Journal of Biological Chemistry, 277, 37280-37291 (2002)
Susceptibility to gastric cancer and polymorphisms of insertion/deletion at the intron 3 of the XRCC4 and VNTR at the promoter region of the XRCC5.
Saadat M, et al.
Pathology Oncology Research, 21, 689-693 (2015)
Blood-based DNA methylation of DNA repair genes in the non-homologous end-joining (NEHJ) pathway in patient with glioma.
Zhou C, et al.
International Journal of Clinical and Experimental Pathology, 8, 9463-9467 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service