Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502546

Sigma-Aldrich

Anti-BAX antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Apoptosis regulator BAX, BAX, BCL2L4, Bcl-2-like protein 4, Bcl2-L-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

Anti-BAX antibody detects endogenous levels of total BAX protein.
The BAX (BCL2 (B-cell lymphoma 2) associated X, apoptosis regulator) gene is mapped to human chromosome 19q13.33. It codes for α-helical globular protein. The protein contains eight amphipathic helices around a hydrophobic helix. Bax is known to be mostly localized in the cytoplasm.

Immunogen

The antiserum was produced against synthesized peptide derived from human Bax.

Immunogen Range: 133-182

Application

Anti-BAX, C-Terminal antibody produced in rabbit has been used in western blotting analysis.

Biochem/physiol Actions

BAX (BCL2 (B-cell lymphoma 2) associated X, apoptosis regulator) is predominantly associated with cellular homeostasis, cell cycle and pathogenesis of cell death particularly in cancer cells. In response to an apoptotic stimulus, Bax brings about mitochondrial dysfunction and apoptosis. Oligomerization of bax promotes mitochondrial outer membrane permeabilization.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and Prognostic Significance of EP300, TP53 and BAX in Clear Cell Renal Cell Carcinoma.
Godlewski J
Anticancer Research, 37(6), 2927-2937 (2017)
Wnt signal transduction pathway and apoptosis: a review.
Pecina-Slaus N
Cancer Cell International, 10:22, 1-5 (2010)
Guo-Liang Zhang et al.
Toxicology and applied pharmacology, 317, 33-40 (2017-01-17)
Zearalenone (ZEA) is an estrogenic mycotoxin mainly produced as a secondary metabolite by numerous species of Fusarium. Previous work showed that ZEA had a negative impact on domestic animals with regard to reproduction. The adverse effects and the mechanisms of
Pro-apoptotic cBid and Bax exhibit distinct membrane remodeling activities: An AFM study.
Unsay JD
Biochimica et Biophysica Acta, 1859(1), 17-27 (2017)
Ping He et al.
International journal of molecular medicine, 45(4), 1261-1269 (2020-03-04)
The authors' previous studies demonstrated that the major renal damage from hepatitis B virus infection is HBx‑induced apoptosis of renal tubular epithelial cells. Cordyceps sinensis is one of the most valuable of traditional Chinese medicines and is extensively used to treat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service