Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502022

Sigma-Aldrich

Anti-NOTCH2 (Cleaved-Val1697) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Motch B, NOTC2, NOTCH2, NTC2, Neurogenic locus notch homolog protein 2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 83 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Val1697)

Gene Information

human ... NOTCH2(4853)

General description

Anti-NOTCH2 (Cleaved-Val1697) antibody detects endogenous levels of fragment of activated NOTCH2 (Cleaved-Val1697) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human NOTCH2.

Immunogen Range: 1678-1727

Biochem/physiol Actions

The NOTCH2 gene is known to be involved in the proliferation, cell cycle progression and colony formation of cancer cells. Upregulation of the gene is observed in hepatocellular carcinoma and salivary adenoid cystic carcinoma. Deletion of NOTCH2 increases the sensitivity of the cancer cells to chemotherapeutic agents such as 5-fluorouracil. NOTCH2 plays an important role in the regulation of liver development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Domenica Giannandrea et al.
Haematologica, 107(9), 2183-2194 (2022-03-11)
Multiple myeloma (MM) is an incurable hematologic neoplasm, whose poor prognosis is deeply affected by the propensity of tumor cells to localize in the bone marrow (BM) and induce the protumorigenic activity of normal BM cells, leading to events associated
Michela Colombo et al.
Haematologica, 105(7), 1925-1936 (2019-10-05)
Multiple myeloma is still incurable due to an intrinsic aggressiveness or, more frequently, to the interactions of malignant plasma cells with the bone marrow (BM) microenvironment. Myeloma cells educate BM cells to support neoplastic cell growth, survival, acquisition of drug
Giuseppina Catanzaro et al.
International journal of molecular sciences, 18(12) (2017-12-21)
The mechanisms by which microRNAs control pediatric high-grade gliomas (pHGGs) have yet to be fully elucidated. Our studies of patient-derived pHGG tissues and of the pHGG cell line KNS42 revealed down-regulation in these tumors of three microRNAs, specifically miR-107, miR-181c
Kai Dai et al.
Frontiers in immunology, 8, 320-320 (2017-04-07)
The mechanisms by which tumor-responsive CD8+ T cells are regulated are important for understanding the tumor immunity and for developing new therapeutic strategies. In current study, we identified the expression of 1810011o10 Rik, which is the homolog of human thyroid
Wen-Rui Wu et al.
Oncology reports, 36(1), 181-188 (2016-05-26)
The Notch pathway plays an important role in both stem cell biology and cancer. Notch2 was reported to be upregulated in human hepatocellular carcinoma (HCC) tissues. However, the biological function of Notch2 in human HCC cells has not yet been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service