Skip to Content
Merck
All Photos(1)

Documents

SAB4501821

Sigma-Aldrich

Anti-TAU antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MAPT, MTBT1, Microtubule-associated protein tau, Neurofibrillary tangle protein, Paired helical filament-tau

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 78 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)

General description

Anti-TAU antibody detects endogenous levels of total TAU protein.
MAPT (microtubule associated protein tau) is located on human chromosome 17q21.3. It is expressed in neurons but is most prominent in axons.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tau.

Immunogen Range: 501-550

Biochem/physiol Actions

Removal of MAPT results in developmental delay and learning disability. It participates in the pathology of Alzheimer′s disease (AD). It helps in the assembly and maintenance of microtubule structure.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youssra K Al-Hilaly et al.
FEBS letters, 594(5), 944-950 (2019-11-14)
The constituent paired helical filaments (PHFs) in neurofibrillary tangles are insoluble intracellular deposits central to the development of Alzheimer's disease (AD) and other tauopathies. Full-length tau requires the addition of anionic cofactors such as heparin to enhance assembly. We have
Youssra K Al-Hilaly et al.
Methods in molecular biology (Clifton, N.J.), 2551, 163-188 (2022-11-01)
Tau is a natively unfolded protein that contributes to the stability of microtubules. Under pathological conditions such as Alzheimer's disease (AD), tau protein misfolds and self-assembles to form paired helical filaments (PHFs) and straight filaments (SFs). Full-length tau protein assembles
Robert Prucek et al.
Scientific reports, 11(1), 6240-6240 (2021-03-20)
Targeted and effective therapy of diseases demands utilization of rapid methods of identification of the given markers. Surface enhanced Raman spectroscopy (SERS) in conjunction with streptavidin-biotin complex is a promising alternative to culture or PCR based methods used for such
Microdeletion encompassing MAPT at chromosome 17q21.3 is associated with developmental delay and learning disability
Shaw-Smith C
Nature Genetics, 38(9), 1032-1037 (2006)
Linkage disequilibrium and association of MAPT H1 in Parkinson disease
Skipper L, et al.
American Journal of Human Genetics, 75(4), 669-677 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service