Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501464

Sigma-Aldrich

Anti-HSP60 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

60 kDa chaperonin, CH60, CPN60, HSP-60, Heat shock protein 60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPD1(3329)

General description

Anti-HSP60 antibody detects endogenous levels of total HSP60 protein.
Hsp60 (heat shock protein family D member 1) is a mitochondrial chaperone, located in the endoplasmic reticulum (ER). It is mapped to human chromosome 2q33.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human HSP60.

Immunogen Range: 511-560

Application

Anti-HSP60, C-Terminal antibody has been used in SDS/PAGE and western blot analysis.

Biochem/physiol Actions

Hsp60 (heat shock protein family D member 1) helps to gather proteins that are imported into mitochondrial matrix. Overexpression of Hsp60 results in Behcet′s Disease (BD),Crohn′s disease, juvenile dermatomyositis (JDM), and juvenile idiopathic arthritis (JIA) and rheumatoid arthritis (RA).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Barrera et al.
FEBS letters, 590(19), 3309-3322 (2016-09-03)
Remodeling of crista junctions (CJs) is observed in numerous human disorders and during apoptosis. The functional interplay of OPA1 and MIC60, two key players in this context, is unclear. We show that OPA1 modulates cristae morphology but is dispensable for
Olivia Harding et al.
Molecular cell, 83(17), 3188-3204 (2023-09-09)
Failure to clear damaged mitochondria via mitophagy disrupts physiological function and may initiate damage signaling via inflammatory cascades, although how these pathways intersect remains unclear. We discovered that nuclear factor kappa B (NF-κB) essential regulator NF-κB effector molecule (NEMO) is
Anti-breast tumor activity of Eclipta extract in-vitro and in-vivo: novel evidence of endoplasmic reticulum specific localization of Hsp60 during apoptosis
Arya RK, et al.
Scientific Reports (2015)
OPA1 functionally interacts with MIC60 but is dispensable for crista junction formation
Barrera M, et al.
Febs Letters, 590(19), 3309-3322 (2016)
HSP60: a double edge sword in autoimmunity
Landstein D, et al.
Oncotarget, 6(32), 32299-32300 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service