Skip to Content
Merck
All Photos(2)

Documents

SAB4501396

Sigma-Aldrich

Anti-ACC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ACAC, ACACA, ACC-α, ACCA, Acetyl-CoA carboxylase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 265 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACACA(31)

General description

Anti-ACC1 Antibody detects endogenous levels of total ACC1 protein.
Acetyl-CoA carboxylase 1 (ACC1) is expressed in adipose tissue, liver, and lactating mammary gland. It is a cytosolic enzyme. The ACC1 gene is mapped to human chromosome 17q12. It comprises biotin carboxylase (BC), carboxyl transferase (CT), and biotin carboxyl carrier protein (BCCP) domains. The BC and CT domains are bridged together through an interaction domain (BT) and a non-catalytic central domain region (CD). The ACC1 gene encompasses three distinct promoter (PI, PII, and PIII) regions.

Immunogen

The antiserum was produced against synthesized peptide derived from human ACC1.

Immunogen Range: 46-95

Application

Anti-ACC1 antibody produced in rabbit has been used in immunoblotting at a dilution 1:500 and immunohistochemistry (1:50 dilution).

Biochem/physiol Actions

Acetyl-CoA carboxylase 1 (ACC1) mediates the carboxylation of acetyl-CoA to form malonyl-CoA in an ATP-dependent manner. It plays a key role in lipogenesis, and its inhibition is regarded as one of the ways to target fatty acid synthesis, especially in metabolic disorders and metabolic syndromes. Haploinsufficiency of ACC1 gene impacts regular fatty acid metabolism. This, in turn, may lead to pathologies associated with infantile encephalitic illness and seizures.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Tonini et al.
Nutrients, 13(6) (2021-07-03)
Bisphenol A (BPA) is an organic chemical compound widely used for manufacturing plastics. BPA exposure originates principally from the diet, but it can also originate from dermal contact. In over 90% of individuals, including pregnant women, BPA is detectable in
Prenatal Exposure to BPA: The Effects on Hepatic Lipid Metabolism in Male and Female Rat Fetuses
Tonini C, et al.
Nutrients null
Moritz Hunkeler et al.
Nature, 558(7710), 470-474 (2018-06-15)
Acetyl-CoA carboxylase catalyses the ATP-dependent carboxylation of acetyl-CoA, a rate-limiting step in fatty acid biosynthesis1,2. Eukaryotic acetyl-CoA carboxylases are large, homodimeric multienzymes. Human acetyl-CoA carboxylase occurs in two isoforms: the metabolic, cytosolic ACC1, and ACC2, which is anchored to the
Hui-Xuan Wu et al.
Endocrine, 73(1), 37-46 (2021-03-22)
17q12 Deletion Syndrome is heterogeneous and the reasons remain unclear. We clarified the clinical characteristics of adulthood diabetes onset 17q12 deletion syndrome and investigated the unclear phenotype-genotype correlation. We collected the clinical history and laboratory results of a family with
Krishna B Singh et al.
Molecular cancer therapeutics, 18(10), 1800-1810 (2019-08-10)
Increased de novo synthesis of fatty acids is implicated in the pathogenesis of human prostate cancer, but a safe and effective clinical inhibitor of this metabolic pathway is still lacking. We have shown previously that leelamine (LLM) suppresses transcriptional activity

Articles

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service