Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300124

Sigma-Aldrich

Anti-phospho-JAK2 (pTyr1007) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JTK10 antibody produced in rabbit, Anti-Janus kinase 2 (a protein tyrosine kinase) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~125 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(K-E-YP-Y-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1007)

Gene Information

human ... JAK2(3717)

General description

Janus kinase 2(JAK2) mapped on human chromosome 9p24, encodes a tyrosine kinase. The encoded protein is a member of the Jak family of protein. JAK2 is ubiquitously expressed. This protein contains a catalytic domain and protein–protein interaction domain.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 1007 (K-E-Y(p)-Y-K), according to the protein JAK2.

Biochem/physiol Actions

Janus kinase 2(JAK2) plays an important role in the activities of cytokines receptors. Overexpression of JAK2 results in neuroblastoma. JAK2 mutations promote primary resistance to PD-1 (anti-programmed death protein 1) blockade therapy. In addition, mutations in the gene has also been observed in polycythaemia vera patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Plays a role in leptin signaling and control of body weight.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A TEL-JAK2 Fusion Protein with Constitutive Kinase Activity in Human Leukemia
Lacronique V
Science, 278, 1309-1312 (1997)
Primary Resistance to PD-1 Blockade Mediated by JAK1/2 Mutations.
Shin DS
Cancer Discovery, 7, 188-201 (2017)
Jak2 Is Essential for Signaling through a Variety of Cytokine Receptors
Parganas E
Cell, 93, 385-395 (1998)
Hodgkin's lymphoma cell lines are characterized by frequent aberrations on chromosomes 2p and 9p including REL and JAK2.
Joos S
International Journal of Cancer. Journal International Du Cancer, 103, 489-495 (2003)
A unique clonal JAK2 mutation leading to constitutive signalling causes polycythaemia vera
James C
Nature, 434, 1144-1148 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service